Sartorius Entris® II Advanced Toploading Balances
Innovative features include real-time level support, CalAuditTrail, and 13 built-in applications
  • Internal electronic level sensor provides built-in real-time level support
  • CalAuditTrail provide automatic documentation of calibration, leveling, and date-and-time stamps
  • Hybrid screen combines LED and graphic touch display to improve functionality
  • Optional isoCAL internal calibration saves you time and money for accurate weighing results
  • Parts offer a high chemical resistance so there is no need to worry over spills
  • Easily transfer data directly to a PC or USB stick with multiple connectivity options
Sartorius Entris® II Advanced precision toploading balances offer higher weighing capacities than their analytical counterparts. The units are constructed of chemical-resistant polybutylene terephthalate, stainless steel, and glass to provide enhanced durability. In addition to high capacity and durability, the balances have advanced features that include dosing, counting, percentage weighing, mixing, totalizing, animal weighing, free factor, density determination, statistics, peakhold, checkweighing, mass unit conversion, and pipette smart test. They enable multiple connectivity options to external devices including RS-232 and USB Type C which offers quick and easy data transfer to a USB stick, PC, or printer.
The Entris® II Advanced line offers enhanced features to ensure accuracy of your measurements. Built-in real-time level support with an internal electronic level sensor continuously monitors proper leveling, provides alarm messaging when your balance isn’t level, and offers interactive user guidance. Integrated protection systems feature three configurable levels to determine valid weighing data and ensure only valid data is transferred to external devices to enhance reliability and accuracy of results. CalAuditTrail provides constant automatic documentation of calibration and levelling processes, and date-and-time stamp.
$2,665.47 - $7,097.20CAD / Each
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Sartorius BCA223-1S Entris® II Advanced Toploading Balance, 220 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197667 Mfr # BCA223-1S
COMPARE
External 220 1
$3,195.34CAD / Each
Sartorius BCA223I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 220 g x 1mg; 100 to 240 VAC, 50/60 Hz 1197668 Mfr # BCA223I-1S
COMPARE
Internal 220 1
$3,741.28CAD / Each
Sartorius BCA323-1S Entris® II Advanced Toploading Balance, 320 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197669 Mfr # BCA323-1S
COMPARE
External 320 1
$3,564.66CAD / Each
Sartorius BCA323I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 320 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197670 Mfr # BCA323I-1S
COMPARE
Internal 320 1
$4,078.48CAD / Each
Sartorius BCA423-1S Entris® II Advanced Toploading Balance, 420 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197671 Mfr # BCA423-1S
COMPARE
External 420 1
$3,901.85CAD / Each
Sartorius BCA423I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 420 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197672 Mfr # BCA423I-1S
COMPARE
Internal 420 1
$4,447.80CAD / Each
Sartorius BCA623-1S Entris® II Advanced Toploading Balance, 620 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197673 Mfr # BCA623-1S
COMPARE
External 620 1
$4,801.04CAD / Each
Sartorius BCA623I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 620 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197674 Mfr # BCA623I-1S
COMPARE
Internal 620 1
$5,330.93CAD / Each
Sartorius BCA822-1S Entris® II Advanced Toploading Balance, 820 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197675 Mfr # BCA822-1S
COMPARE
External 820 10
$2,842.10CAD / Each
Sartorius BCA822I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 820 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197676 Mfr # BCA822I-1S
COMPARE
Internal 820 10
$3,388.03CAD / Each
Sartorius BCA1203I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 1200 g x 1 mg; 100 to 240 VAC, 50/60 Hz 1197677 Mfr # BCA1203I-1S
COMPARE
Internal 1200 1
$7,097.20CAD / Each
Sartorius BCA1202-1S Entris® II Advanced Toploading Balance, 1200 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197678 Mfr # BCA1202-1S
COMPARE
External 1200 10
$3,195.34CAD / Each
Sartorius BCA1202I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 1200 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197679 Mfr # BCA1202I-1S
COMPARE
Internal 1200 10
$3,741.28CAD / Each
Sartorius BCA2201-1S Entris® II Advanced Toploading Balance, 2200 g x 0.1 g; 100 to 240 VAC, 50/60 Hz 1197680 Mfr # BCA2201-1S
COMPARE
External 2200 100
$2,665.47CAD / Each
Sartorius BCA2201I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 2200 g x 0.1 g; 100 to 240 VAC, 50/60 Hz 1197681 Mfr # BCA2201I-1S
COMPARE
Internal 2200 100
$3,195.34CAD / Each
Sartorius BCA2202-1S Entris® II Advanced Toploading Balance, 2200 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197682 Mfr # BCA2202-1S
COMPARE
External 2200 10
$3,564.66CAD / Each
Sartorius BCA2202I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 2200 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197683 Mfr # BCA2202I-1S
COMPARE
Internal 2200 10
$4,078.48CAD / Each
Sartorius BCA3202-1S Entris® II Advanced Toploading Balance, 3200 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197684 Mfr # BCA3202-1S
COMPARE
External 3200 10
$3,901.85CAD / Each
Sartorius BCA3202I-1S Entris® II Advanced Toploading Balance with Internal Calibration, 3200 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197685 Mfr # BCA3202I-1S
COMPARE
Internal 3200 10
$4,447.80CAD / Each
Sartorius BCA4202-1S Entris® II Advanced Toploading Balance, 4200 g x 10 mg; 100 to 240 VAC, 50/60 Hz 1197686 Mfr # BCA4202-1S
COMPARE
External 4200 10
$4,078.48CAD / Each
Showing 1 - 20 of 31