Sartorius Arium® Comfort II Water Purification Systems
With space saving twin technology, the Arium® Comfort I delivers ASTM Type 1 ultrapure water and Type 2 pure water all in one system
  • Use of innovative bag technology, eliminates costly tank cleaning
  • Optimized water consumption is automatic with iJust software
  • Easy to use display with touch function and intuitive menu
  • Favorites function with direct access for recurring volumes
This system contains state-of-the-art reverse osmosis technology and a unique cartridge specifically for the production of high ultrapure water quality. With the optionally integrated TOC monitor, its compact design, the flexible display, and the SD card slot, the Arium® Comfort II is the ideal choice for demanding laboratory applications. Compared to conventional water systems, the Arium® Comfort II optimizes water consumption using the integrated iJust control unit. Directly connect the systems to the potable water and purify the water in two stages. In the first stage, an Arium® Comfort II system features built-in EDI technology (electrodeionization) and produces Type 2 ultrapure water. In the second stage, the system is identical to an Arium® Comfort I and produces ASTM Type 1 ultrapure water.
$26,360.66 - $31,459.37CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description ASTM Water Grade With UV Lamp
Availability Pricing
Sartorius Arium® Comfort II Wall-Mounted System, 5 L/h, without TOC Monitor and UV Lamp 9929358 Mfr # H2O-II-1-B
COMPARE
Arium® Comfort II Wall-Mounted System, 5 L/h, without TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water No
$26,360.66CAD / Each
Sartorius Arium® Comfort II Bench-Top System, 5 L/h, without TOC Monitor and UV Lamp 9929359 Mfr # H2O-II-1-T
COMPARE
Arium® Comfort II Bench-Top System, 5 L/h, without TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water No
$26,360.66CAD / Each
Sartorius Arium® Comfort II Wall-Mounted System, 5 L/h, with TOC Monitor and UV Lamp 9929360 Mfr # H2O-II-1-TOC-B
COMPARE
Arium® Comfort II Wall-Mounted System, 5 L/h, with TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$28,111.43CAD / Each
Sartorius Arium® Comfort II Bench-Top System, 5 L/h, with TOC Monitor and UV Lamp 9929361 Mfr # H2O-II-1-TOC-T
COMPARE
Arium® Comfort II Bench-Top System, 5 L/h, with TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$28,111.43CAD / Each
Sartorius Arium® Comfort II Wall-Mounted System, 5 L/h, with UV Lamp 9929362 Mfr # H2O-II-1-UV-B
COMPARE
Arium® Comfort II Wall-Mounted System, 5 L/h, with UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$26,790.09CAD / Each
Sartorius Arium® Comfort II Bench-Top System, 5 L/h, with UV Lamp 9929363 Mfr # H2O-II-1-UV-T
COMPARE
Arium® Comfort II Bench-Top System, 5 L/h, with UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$26,790.09CAD / Each
Sartorius Arium® Comfort II Wall-Mounted System, 10 L/h, without TOC Monitor and UV Lamp 9929364 Mfr # H2O-II-2-B
COMPARE
Arium® Comfort II Wall-Mounted System, 10 L/h, without TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water No
$28,772.09CAD / Each
Sartorius Arium® Comfort II Bench-Top System, 10 L/h, without TOC Monitor and UV Lamp 9929365 Mfr # H2O-II-2-T
COMPARE
Arium® Comfort II Bench-Top System, 10 L/h, without TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water No
$28,772.09CAD / Each
Sartorius Arium® Comfort II Wall-Mounted System, 10 L/h, with TOC Monitor and UV Lamp 9929366 Mfr # H2O-II-2-TOC-B
COMPARE
Arium® Comfort II Wall-Mounted System, 10 L/h, with TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$31,459.37CAD / Each
Sartorius Arium®Comfort II Bench-Top System, 10 L/h, with TOC Monitor and UV Lamp 9929367 Mfr # H2O-II-2-TOC-T
COMPARE
Arium®Comfort II Bench-Top System, 10 L/h, with TOC Monitor and UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$31,459.37CAD / Each
Sartorius Arium® Comfort II Wall-Mounted System, 10 L/h, with UV Lamp 9929368 Mfr # H2O-II-2-UV-B
COMPARE
Arium® Comfort II Wall-Mounted System, 10 L/h, with UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$29,085.91CAD / Each
Sartorius Arium® Comfort II Bench-Top System, 10 L/h, with UV Lamp 9929369 Mfr # H2O-II-2-UV-T
COMPARE
Arium® Comfort II Bench-Top System, 10 L/h, with UV Lamp ASTM Type 1 ultrapure water and Type 2 pure water Yes
$29,069.40CAD / Each
Showing 1- 12 of 12