Sartorius Arium® Comfort I Water Purification Systems
With space saving twin technology, the Arium® Comfort I delivers ASTM Type 1 ultrapure water and Type 3 pure water all in one system
  • Use of innovative bag technology, eliminates costly tank cleaning
  • Optimized water consumption is automatic with iJust software
  • Easy to use display with touch function and intuitive menu
  • Favorites function with direct access for recurring volumes
This system contains state-of-the-art reverse osmosis technology and a unique cartridge specifically for the production of high ultrapure water quality. With the optionally integrated TOC monitor, its compact design, the flexible display, and the SD card slot, the Arium® Comfort I is the ideal choice for demanding laboratory applications. Compared to conventional water systems, the Arium® Comfort I optimizes water consumption using the integrated iJust control unit. Directly connect the systems to the potable water and purify the water in two stages. In the first stage, an Arium® Comfort I system produces Type 3 pure water (reverse osmosis water), and in the second stage, ASTM Type 1 ultrapure water. The pure water is stored in the enclosed Arium® Bagtank system. This guarantees optimal storage and protects against secondary contamination. Time-consuming tank cleaning intervals are eliminated thanks to the interchangeable bag. With the new favorites function it is possible to save recurring volumes and retrieve them as required by direct access.
$14,600.76 - $20,001.73CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description ASTM Water Grade With UV Lamp
Availability Pricing
Sartorius Arium® Comfort I Wall-Mounted System, 8 L/h, without TOC Monitor and UV Lamp 9929346 Mfr # H2O-I-1-B
COMPARE
Arium® Comfort I Wall-Mounted System, 8 L/h, without TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure No
$14,600.76CAD / Each
Sartorius Arium® Comfort I Bench-Top System, 8 L/h, without TOC Monitor and UV Lamp 9929347 Mfr # H2O-I-1-T
COMPARE
Arium® Comfort I Bench-Top System, 8 L/h, without TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure No
$14,600.76CAD / Each
Sartorius Arium® Comfort I Wall-Mounted System, 8 L/h, with TOC Monitor and UV Lamp 9929348 Mfr # H2O-I-1-TOC-B
COMPARE
Arium® Comfort I Wall-Mounted System, 8 L/h, with TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure Yes
$17,238.49CAD / Each
Sartorius Arium® Comfort I Bench-Top System, 8 L/h, with TOC Monitor and UV Lamp 9929349 Mfr # H2O-I-1-TOC-T
COMPARE
Arium® Comfort I Bench-Top System, 8 L/h, with TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure Yes
$17,238.49CAD / Each
Sartorius Arium® Comfort I Wall-Mounted System, 8 L/h, with UV Lamp 9929350 Mfr # H2O-I-1-UV-B
COMPARE
Arium® Comfort I Wall-Mounted System, 8 L/h, with UV Lamp Type 1 ultrapure and Type 3 pure Yes
$15,330.80CAD / Each
Sartorius Arium® Comfort I Bench-Top System, 8 L/h, with UV Lamp 9929351 Mfr # H2O-I-1-UV-T
COMPARE
Arium® Comfort I Bench-Top System, 8 L/h, with UV Lamp Type 1 ultrapure and Type 3 pure Yes
$15,330.80CAD / Each
Sartorius Arium® Comfort I Wall-Mounted System, 16 L/h, without TOC Monitor and UV Lamp 9929352 Mfr # H2O-I-2-B
COMPARE
Arium® Comfort I Wall-Mounted System, 16 L/h, without TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure No
$17,218.67CAD / Each
Sartorius Arium® Comfort I Bench-Top System, 16 L/h, without TOC Monitor and UV Lamp 9929353 Mfr # H2O-I-2-T
COMPARE
Arium® Comfort I Bench-Top System, 16 L/h, without TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure No
$17,218.67CAD / Each
Sartorius Arium® Comfort I Wall-Mounted System, 16 L/h, with TOC Monitor and UV Lamp 9929354 Mfr # H2O-I-2-TOC-B
COMPARE
Arium® Comfort I Wall-Mounted System, 16 L/h, with TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure Yes
$20,001.73CAD / Each
Sartorius Arium® Comfort I Bench-Top System, 16 L/h, with TOC Monitor and UV Lamp 9929355 Mfr # H2O-I-2-TOC-T
COMPARE
Arium® Comfort I Bench-Top System, 16 L/h, with TOC Monitor and UV Lamp Type 1 ultrapure and Type 3 pure Yes
$20,001.73CAD / Each
Sartorius Arium® Comfort I Wall-Mounted System, 16 L/h, with UV Lamp 9929356 Mfr # H2O-I-2-UV-B
COMPARE
Arium® Comfort I Wall-Mounted System, 16 L/h, with UV Lamp Type 1 ultrapure and Type 3 pure Yes
$17,945.40CAD / Each
Sartorius Arium® Comfort I Bench-Top System, 16 L/h, with UV Lamp 9929357 Mfr # H2O-I-2-UV-T
COMPARE
Arium® Comfort I Bench-Top System, 16 L/h, with UV Lamp Type 1 ultrapure and Type 3 pure Yes
$17,945.40CAD / Each
Showing 1- 12 of 12