Sartorius Arium® Advance Reverse Osmosis (RO) Water Purification Systems
The innovative iJust technology automatically optimizes water production
  • Achieves the highest quality ASTM Type III reverse osmosis water
  • Easy-to-use touch display with intuitive menu navigation
  • Must be used with Arium Bagtank system—order separately
Directly connect the Arium advance reverse osmosis water purification systems to potable feed water source to produce ASTM Type III water. Type III water is ideal for general laboratory applications such as rinsing glass and laboratory containers, feed water for laboratory equipment, or feed water for ultrapure water systems. The innovative iJust software ensures the highest water quality and optimizes water consumption, guaranteeing a longer life of the downstream ultrapure water systems.
Easily program and run the system using four function keys. This simple-to-navigate control panel is easy to use even while wearing gloves. View time, date, and working environment (conductivity while filling, fill volume, and flow diagram) during operating mode.
Store pure water produced by the Advance RO in the enclosed Arium Bagtank system (order separately). This innovative bag system protects purified water against secondary contamination while eliminating the time-consuming tank cleaning intervals. Dispense water directly for further use in ported equipment or manually using an optional remote dispenser.
Use the Ethernet interface to connect an Arium Smart Station for remote dispensing of pure water from the Bagtank directly at the point of use. The Arium Exchange interface enables communication with an Arium ultrapure water system to stop dispensing water if the fill level of the Bagtank falls below the minimum (<2 L).
$12,139.78 - $15,393.57CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description Max Production (L/hour) Special Features
Availability Pricing
Sartorius Arium® Advance Reverse Osmosis Wall-Mount Water Purification System, 8 L/hr 9929384 Mfr # H2O-RO-1-B
COMPARE
Arium® Advance Reverse Osmosis Wall-Mount Water Purification System, 8 L/hr 8 Wall mount
$12,139.78CAD / Each
Sartorius Arium® Advance Reverse Osmosis Benchtop Water Purification System, 8 L/hr 9929385 Mfr # H2O-RO-1-T
COMPARE
Arium® Advance Reverse Osmosis Benchtop Water Purification System, 8 L/hr 8 Benchtop
$12,139.78CAD / Each
Sartorius Arium® Advance Reverse Osmosis Wall-Mount Water Purification System, 16 L/hr 9929386 Mfr # H2O-RO-2-B
COMPARE
Arium® Advance Reverse Osmosis Wall-Mount Water Purification System, 16 L/hr 16 Wall mount
$13,684.09CAD / Each
Sartorius Arium® Advance Reverse Osmosis Benchtop Water Purification System, 16 L/hr 9929387 Mfr # H2O-RO-2-T
COMPARE
Arium® Advance Reverse Osmosis Benchtop Water Purification System, 16 L/hr 16 Benchtop
$13,684.09CAD / Each
Sartorius Arium® Advance Reverse Osmosis Wall-Mount Water Purification System, 24 L/hr 9929388 Mfr # H2O-RO-3-B
COMPARE
Arium® Advance Reverse Osmosis Wall-Mount Water Purification System, 24 L/hr 24 Wall mount
$15,393.57CAD / Each
Sartorius Arium® Advance Reverse Osmosis Benchtop Water Purification System, 24 L/hr 9929389 Mfr # H2O-RO-3-T
COMPARE
Arium® Advance Reverse Osmosis Benchtop Water Purification System, 24 L/hr 24 Benchtop
$15,393.57CAD / Each