Saint-Gobain PharmaFluor ® Compression Fitting, Elbow Union, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
$89.88 - $136.29CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem Availability Pricing
Saint Gobain PharmaFluor ® Compression Fitting, Elbow Union, PFA, 1/2" OD 5014464 Mfr # G1077025
COMPARE
$102.40CAD / Each
PharmaFluor ® Compression Fitting, Elbow Union, PFA, 1/4" OD 5014465 Mfr # G1077023
COMPARE
$89.88CAD / Each
Saint Gobain PharmaFluor ® Compression Fitting, Elbow Union, PFA, 3/4" OD 5014466 Mfr # G1077026
COMPARE
$136.29CAD / Each
Saint Gobain PharmaFluor ® Compression Fitting, Elbow Union, PFA, 3/8" OD 5014467 Mfr # G1077024
COMPARE
$93.56CAD / Each