QLA Sotax-Compatible Baskets
QLA Baskets dissolution testing with your Sotax apparatus
  • Made of 316 electropolished stainless steel All baskets are laser marked with unique serial numbers
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Sotax test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
$238.32 - $1,100.62CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Clip Style Basket for Sotax, 10 Mesh, 316 SS; Serialized 4130134 Mfr # BSK010-ST
COMPARE
316 stainless steel 10 Clip Style Basket for Sotax, 10 Mesh, 316 SS; Serialized
$292.84CAD / Each
QLA Clip Style Basket for Sotax, 20 Mesh, 316 SS; Serialized 4130142 Mfr # BSK020-ST
COMPARE
316 stainless steel 20 Clip Style Basket for Sotax, 20 Mesh, 316 SS; Serialized
$297.99CAD / Each
QLA Clip Style Basket for Sotax, 40 Mesh USP, 316 SS; Serialized 4130152 Mfr # BSK040-ST
COMPARE
316 stainless steel 40 Clip Style Basket for Sotax, 40 Mesh USP, 316 SS; Serialized
$238.32CAD / Each
QLA Clip Style Basket for Sotax, 80 Mesh, 316 SS; Serialized 4130157 Mfr # BSK080-ST
COMPARE
316 stainless steel 80 Clip Style Basket for Sotax, 80 Mesh, 316 SS; Serialized
$579.04CAD / Each
QLA Clip Style Basket for Sotax, 100 Mesh, 316 SS; Serialized 4130162 Mfr # BSK100-ST
COMPARE
316 stainless steel 100 Clip Style Basket for Sotax, 100 Mesh, 316 SS; Serialized
$889.93CAD / Each
QLA Clip Style Basket for Sotax, 140 Mesh, 316 SS; Serialized 4130165 Mfr # BSK140-ST
COMPARE
316 stainless steel 140 Clip Style Basket for Sotax, 140 Mesh, 316 SS; Serialized
$769.11CAD / Each
QLA Suppository Basket for Sotax; Plastic 4130198 Mfr # BSKSUP-ST
COMPARE
Plastic Suppository Basket for Sotax; Plastic
$340.35CAD / Each
QLA Clip Style Basket for Sotax, 100 Mesh with 20 Mesh Stability Lining, 316SS; Serialized 4130845 Mfr # CUSBSK-20/100ST
COMPARE
316 stainless steel 100 (plus 20 mesh stability lining) Clip Style Basket for Sotax, 100 Mesh with 20 Mesh Stability Lining, 316SS; Serialized
$1,100.62CAD / Each
QLA Clip Style Basket for Sotax, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130852 Mfr # CUSBSK-20/150ST
COMPARE
316 stainless steel 150 (plus 20 mesh stability lining) Clip Style Basket for Sotax, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
$1,100.62CAD / Each
QLA Custom Basket for Sotax Automated System, 15 Mesh; 75% open area 4130857 Mfr # CUSTBSK-15ST
COMPARE
316 stainless steel 15 (75% open area) Custom Basket for Sotax Automated System, 15 Mesh; 75% open area
$342.19CAD / Each
QLA Custom Basket for Sotax Automated System, 28 Mesh; 52% open area 4130858 Mfr # CUSTBSK-28ST
COMPARE
316 stainless steel 28 (52% open area) Custom Basket for Sotax Automated System, 28 Mesh; 52% open area
$342.19CAD / Each
Showing 1- 11 of 11