QLA Shafts Compatible with Agilent, VanKel, and Varian Dissolution Apparatus
QLA Shafts for dissolution testing with your Agilent/VanKel/Varian apparatus
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
These QLA shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Agilent/VanKel/Varian test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. Shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
$307.57 - $520.84CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Length (in) Material Description
Availability Pricing
QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian V-Series Dissolution Apparatus, 14½''L, 316 SS; Serialized 4130177 Mfr # BSKSHT-13
COMPARE
14.5 316 stainless steel Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian V-Series Dissolution Apparatus, 14½''L, 316 SS; Serialized
$307.57CAD / Each
QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 15"L, 316 SS; Serialized 4130178 Mfr # BSKSHT-15
COMPARE
15 316 stainless steel Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 15"L, 316 SS; Serialized
$311.25CAD / Each
QLA Spring Clip Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 15''L, 316 SS; Serialized 4130179 Mfr # BSKSHT-15C
COMPARE
15 316 stainless steel Spring Clip Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 15''L, 316 SS; Serialized
$321.94CAD / Each
QLA Push-On Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 15''L, 316 SS; Serialized 4130180 Mfr # BSKSHT-15CO
COMPARE
15 316 stainless steel Push-On Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 15''L, 316 SS; Serialized
$321.94CAD / Each
QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 16"L, 316 SS; Serialized 4130183 Mfr # BSKSHT-16VK
COMPARE
16 316 stainless steel Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 16"L, 316 SS; Serialized
$393.76CAD / Each
QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 19"L, 316 SS; Serialized 4130184 Mfr # BSKSHT-19
COMPARE
19 316 stainless steel Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 19"L, 316 SS; Serialized
$340.35CAD / Each
QLA Spring Clip Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 19"L, 316 SS; Serialized 4130185 Mfr # BSKSHT-19C
COMPARE
19 316 stainless steel Spring Clip Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 19"L, 316 SS; Serialized
$345.51CAD / Each
QLA Push-On Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 19"L, 316 SS; Serialized 4130186 Mfr # BSKSHT-19CO
COMPARE
19 316 stainless steel Push-On Style Conical Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 19"L, 316 SS; Serialized
$348.83CAD / Each
QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 21"L, 316 SS; Serialized 4130188 Mfr # BSKSHT-21
COMPARE
21 316 stainless steel Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 21"L, 316 SS; Serialized
$433.18CAD / Each
QLA Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 24"L, 316 SS; Serialized 4130189 Mfr # BSKSHT-24
COMPARE
24 316 stainless steel Spring Clip Style Basket Shaft Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 24"L, 316 SS; Serialized
$520.84CAD / Each
Showing 1- 10 of 10