QLA Porous Cannula Filters for Pharmatest Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds and provide a good flow rate.
$183.44 - $1,989.08CAD / Pkg of 1000
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron) Description
Availability Pricing
QLA Porous Filters, 1 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK 4130882 Mfr # FIL001-PT
COMPARE
1 Porous Filters, 1 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK
$1,989.08CAD / Pkg of 1000
QLA Porous Filters, 1 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK 4130883 Mfr # FIL001-PT-a
COMPARE
1 Porous Filters, 1 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK
$280.68CAD / Pkg of 100
QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK 4130892 Mfr # FIL004-PT
COMPARE
4 Porous Filters, 4 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK
$1,989.08CAD / Pkg of 1000
QLA Porous Filters, 4 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK 4130893 Mfr # FIL004-PT-a
COMPARE
4 Porous Filters, 4 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK
$280.68CAD / Pkg of 100
QLA Porous Filters, 5 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK 4130894 Mfr # FIL005-PT
COMPARE
5 Porous Filters, 5 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK
$1,989.08CAD / Pkg of 1000
QLA Porous Filters, 5 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK 4130895 Mfr # FIL005-PT-A
COMPARE
5 Porous Filters, 5 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK
$280.68CAD / Pkg of 100
QLA Porous Filters, 10 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK 4130908 Mfr # FIL010-PT
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK
$1,532.33CAD / Pkg of 1000
QLA Porous Filters, 10 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK 4130909 Mfr # FIL010-PT-A
COMPARE
10 Porous Filters, 10 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK
$183.44CAD / Pkg of 100
QLA Porous Filters, 20 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK 4130922 Mfr # FIL020-PT
COMPARE
20 Porous Filters, 20 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK
$1,849.10CAD / Pkg of 1000
QLA Porous Filters, 20 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK 4130923 Mfr # FIL020-PT-A
COMPARE
20 Porous Filters, 20 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK
$220.64CAD / Pkg of 100
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK 4130937 Mfr # FIL035-PT
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK
$1,554.43CAD / Pkg of 1000
QLA Porous Filters, 35 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK 4130938 Mfr # FIL035-PT-A
COMPARE
35 Porous Filters, 35 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK
$191.91CAD / Pkg of 100
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK 4130965 Mfr # FIL070-PT
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 1000/PK
$1,849.10CAD / Pkg of 1000
QLA Porous Filters, 70 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK 4130966 Mfr # FIL070-PT-a
COMPARE
70 Porous Filters, 70 µm, UHMW PE, 1/8" ID, Pharmatest compatible; 100/PK
$238.32CAD / Pkg of 100
Showing 1- 14 of 14