QLA Distek-Compatible Sampling Probes
Achieve consistent sampling with QLA Distek-compatible probes
  • Matching cannula to stopper ensures greater sampling position consistency Made of stainless steel that will not absorb or interfere with active ingredients
For dissolution tests, taking reliable samples is critical. QLA probes and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
$143.51 - $351.77CAD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
QLA Standard Sampling Probe for Distek Baths; 1000 mL 4130220 Mfr # CAN01K-DK
COMPARE
1000 mL vessel Standard Sampling Probe for Distek Baths; 1000 mL
$254.90CAD / Each
QLA Solid Housing Sampling Probe for Distek Baths; 1000 mL 4130221 Mfr # CAN01K-DSH
COMPARE
1000 mL vessel Solid Housing Sampling Probe for Distek Baths; 1000 mL
$246.42CAD / Each
QLA DV Series Sampling Probe Assembly for Distek Baths; 1000 mL 4130222 Mfr # CAN01K-DV
COMPARE
1000 mL vessel DV Series Sampling Probe Assembly for Distek Baths; 1000 mL
$268.89CAD / Each
QLA Standard Sampling Probe for Distek Baths; 2000 mL 4130223 Mfr # CAN02K-DK
COMPARE
2000 mL vessel Standard Sampling Probe for Distek Baths; 2000 mL
$246.42CAD / Each
QLA DV Series Sampling Probe Assembly for Distek Baths; 2000 mL 4130224 Mfr # CAN02K-DV
COMPARE
2000 mL vessel DV Series Sampling Probe Assembly for Distek Baths; 2000 mL
$246.42CAD / Each
QLA DV Series Sampling Probe Assembly for Distek Baths; 500 mL 4130237 Mfr # CAN500-DDV
COMPARE
500 mL vessel DV Series Sampling Probe Assembly for Distek Baths; 500 mL
$351.77CAD / Each
QLA Standard Sampling Probe for Distek Baths; 500 mL 4130239 Mfr # CAN500-DK
COMPARE
500 mL vessel Standard Sampling Probe for Distek Baths; 500 mL
$274.05CAD / Each
QLA Solid Housing Sampling Probe for Distek Baths; 500 mL 4130240 Mfr # CAN500-DSH
COMPARE
500 mL vessel Solid Housing Sampling Probe for Distek Baths; 500 mL
$270.37CAD / Each
QLA DV Series Sampling Probe for Distek Baths; 500 mL 4130241 Mfr # CAN500-DV
COMPARE
500 mL vessel DV Series Sampling Probe for Distek Baths; 500 mL
$265.58CAD / Each
QLA Standard Sampling Probe Bottom for Distek Baths; 500 mL 4130243 Mfr # CAN5BM-DK
COMPARE
500 mL vessel Standard Sampling Probe Bottom for Distek Baths; 500 mL
$160.97CAD / Each
QLA DV Series Sampling Probe Bottom for Distek Baths; 500 mL 4130244 Mfr # CAN5BM-DV
COMPARE
500 mL vessel DV Series Sampling Probe Bottom for Distek Baths; 500 mL
$160.97CAD / Each
QLA Dual DV Series Sampling Probe for Distek Baths; 900 mL 4130254 Mfr # CAN900-DDV
COMPARE
900 mL vessel Dual DV Series Sampling Probe for Distek Baths; 900 mL
$351.77CAD / Each
QLA Standard Sampling Probe for Distek Baths; 900 mL 4130257 Mfr # CAN900-DK
COMPARE
900 mL vessel Standard Sampling Probe for Distek Baths; 900 mL
$250.11CAD / Each
QLA Solid Housing Sampling Probe for Distek Baths; 900 mL 4130258 Mfr # CAN900-DSH
COMPARE
900 mL vessel Solid Housing Sampling Probe for Distek Baths; 900 mL
$250.11CAD / Each
QLA DV Series Sampling Probe for Distek Baths; 900 mL 4130259 Mfr # CAN900-DV
COMPARE
900 mL vessel DV Series Sampling Probe for Distek Baths; 900 mL
$283.63CAD / Each
QLA Standard Sampling Probe Bottom for Distek Baths; 900 mL 4130261 Mfr # CAN9BM-DK
COMPARE
900 mL vessel Standard Sampling Probe Bottom for Distek Baths; 900 mL
$160.97CAD / Each
QLA DV Series Sampling Probe Bottom for Distek Baths; 900 mL 4130262 Mfr # CAN9BM-DV
COMPARE
900 mL vessel DV Series Sampling Probe Bottom for Distek Baths; 900 mL
$160.97CAD / Each
QLA Return Probe for Distek Baths 4130264 Mfr # CANRET-DK
COMPARE
Return Probe for Distek Baths
$173.86CAD / Each
QLA Standard Sampling Probe Top for Distek Baths 4130283 Mfr # CANTOP-DK
COMPARE
Standard Sampling Probe Top for Distek Baths
$166.86CAD / Each
QLA DV Series Sampling Probe Top for Distek Baths 4130284 Mfr # CANTOP-DV
COMPARE
DV Series Sampling Probe Top for Distek Baths
$143.51CAD / Each
Showing 1- 20 of 20