QLA Baskets Compatible with Agilent, VanKel, and Varian Dissolution Apparatus
QLA Baskets for dissolution testing with your Agilent/VanKel/Varian apparatus
  • Made of 316 electropolished stainless steel
  • All baskets are laser marked with unique serial numbers
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Agilent/VanKel/Varian test apparatus. All (except plastic suppository baskets) are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
$231.32 - $1,100.62CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 10 Mesh, 316 SS; Serialized 4130127 Mfr # BSK010-01
COMPARE
316 stainless steel 10 Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 10 Mesh, 316 SS; Serialized
$356.93CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 20 Mesh, 316 SS; Serialized 4130135 Mfr # BSK020-01
COMPARE
316 stainless steel 20 Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 20 Mesh, 316 SS; Serialized
$360.61CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 30 Mesh, 316 SS; Serialized 4130143 Mfr # BSK030-01
COMPARE
316 stainless steel 30 Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 30 Mesh, 316 SS; Serialized
$372.03CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 40 Mesh USP, 316 SS; Serialized 4130145 Mfr # BSK040-01
COMPARE
316 stainless steel 40 Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 40 Mesh USP, 316 SS; Serialized
$231.32CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 Mesh, 316 SS, Serialized 4130158 Mfr # BSK100-01
COMPARE
316 stainless steel 100 Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 Mesh, 316 SS, Serialized
$891.40CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 150 Mesh, 316 SS; Serialized 4130166 Mfr # BSK150-01
COMPARE
316 stainless steel 150 Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 150 Mesh, 316 SS; Serialized
$941.50CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 40 Mesh, PTFE Coated over 316 SS; Serialized 4130174 Mfr # BSKPTV-40
COMPARE
PTFE-coated 316 SS 40 Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 40 Mesh, PTFE Coated over 316 SS; Serialized
$531.16CAD / Each
QLA Suppository Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus; Plastic 4130192 Mfr # BSKSUP-01
COMPARE
Plastic Suppository Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus; Plastic
$340.35CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130846 Mfr # CUSBSK-20/100VK
COMPARE
316 stainless steel 100 (plus 20 mesh stability lining) Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 100 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
$1,100.62CAD / Each
QLA Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized 4130853 Mfr # CUSBSK-20/150VK
COMPARE
316 stainless steel 150 (plus 20 mesh stability lining) Clip Style Basket Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 150 Mesh with 20 Mesh Stability Lining, 316 SS; Serialized
$1,100.62CAD / Each
Showing 1- 10 of 10