PYREX® Distillation Apparatus

General purpose batch distillation apparati allow the addition of reagents via the No. 27 Standard Taper PYREX® stoppered neck
  • Condenser tubulations: 10 mm outer diameter
  • Condenser jacket length: 200 mm
Distillation apparatus is used for the batch production of high purity distilled water.
$774.49 - $899.80CAD / Case of 1

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accessory Type Range
Availability Pricing
Corning 3360-500 Distillation Apparatus, 500 Ml 3451020 Mfr # 3360-500
COMPARE
Distillation Apparatus 500 mL
$774.49CAD / Case of 1
Corning 3360-1L Distillation Apparatus, 1000 Ml 3451022 Mfr # 3360-1L
COMPARE
Distillation Apparatus 1 L
$899.80CAD / Case of 1
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat