Pyrex Desiccators, Glass
Borosilicate glass, knob top
  • Durable and efficient
These desiccators are designed for analytical work. The sturdy knob makes the cover easy to handle and move. Desicctors require desiccator plates, order separately.
Warning: Do not heat or subject to pressure. Not for vacuum applications.
$474.43 - $1,156.61CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Volume (Liters) Plate Size (mm)
Availability Pricing
Pyrex 3081-150 Desiccator, 2.4 L, Glass, 5.5" dia 3454819 Mfr # 3081-150
COMPARE
2.4 140
$474.43CAD / Each
Pyrex 3081-250 Desiccator, 10.5 L, Glass, 9" dia, Clear 3454824 Mfr # 3081-250
COMPARE
10.5 230
$1,156.61CAD / Case of 1