PYREX® Brand 1000 Griffin Low-Form Beakers
Stand up to thermal shock
  • No mystery liquids—extra-large marking spot for easy identification
Glass beakers with spouts are manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength. The 250-mL to 4000-mL beakers have a double graduated metric scale to indicate approximate content. All sizes have an extra large marking spot.
$83.69 - $832.47CAD / Pkg of 1
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex 1000-50 Brand 1000 Griffin Low-Form Beaker, 50 mL, 12/pk 3450204 Mfr # 1000-50
COMPARE
50 10 Glass
$150.29CAD / Pkg of 12
Pyrex 1000-100 Brand 1000 Griffin Low-Form Beaker, 100 mL, 12/pk 3450205 Mfr # 1000-100
COMPARE
100 10 Glass
$160.60CAD / Pkg of 12
Pyrex 1000-150 Brand 1000 Griffin Low-Form Beaker, 150 mL, 12/pk 3450206 Mfr # 1000-150
COMPARE
150 10 Glass
$148.81CAD / Pkg of 12
Pyrex 1000-250 Brand 1000 Griffin Low-Form Beaker, 250 mL, 12/pk 3450207 Mfr # 1000-250
COMPARE
250 25 Glass
$160.60CAD / Pkg of 12
Pyrex 1000-400 Brand 1000 Griffin Low-Form Beaker, 400 mL, 12/pk 3450208 Mfr # 1000-400
COMPARE
400 25 Glass
$179.75CAD / Pkg of 12
Pyrex 1000-600 Brand 1000 Griffin Low-Form Beaker, 600 mL, 6/pk 3450209 Mfr # 1000-600
COMPARE
600 50 Glass
$115.66CAD / Pkg of 6
Pyrex 1000-800 Brand 1000 Griffin Low-Form Beaker, 800 mL, 6/pk 3450210 Mfr # 1000-800
COMPARE
800 50 Glass
$160.60CAD / Pkg of 6
Pyrex 1000-1L Brand 1000 Griffin Low-Form Beaker, 1000 mL, 6/pk 3450211 Mfr # 1000-1L
COMPARE
1000 50 Glass
$187.12CAD / Pkg of 6
Pyrex 1000-2L Brand 1000 Griffin Low-Form Beaker, 2000 mL, 4/pk 3450213 Mfr # 1000-2L
COMPARE
2000 100 Glass
$244.58CAD / Pkg of 4
Pyrex 1000-3L Brand 1000 Griffin Low-Form Beaker, 3000 mL, 1/pk 3450214 Mfr # 1000-3L
COMPARE
3000 125 Glass
$127.45CAD / Pkg of 1
Pyrex 1000-4L Brand 1000 Griffin Low-Form Beaker, 4000 mL, 1/pk 3450215 Mfr # 1000-4L
COMPARE
4000 250 Glass
$235.74CAD / Pkg of 1
Pyrex 1000-20 Brand 1000 Griffin Low-Form Beaker, 20 mL, 48/cs 3450222 Mfr # 1000-20
COMPARE
20 5 Glass
$383.08CAD / Case of 48
Pyrex 1000-30 Brand 1000 Griffin Low-Form Beaker, 30 mL, 48/cs 3450223 Mfr # 1000-30
COMPARE
30 10 Glass
$372.77CAD / Case of 48
Pyrex 1000-50 Brand 1000 Griffin Low-Form Beaker, 50 mL, 48/cs 3450224 Mfr # 1000-50
COMPARE
50 10 Glass
$372.77CAD / Case of 48
Pyrex 1000-100 Brand 1000 Griffin Low-Form Beaker, 100 mL, 48/cs 3450225 Mfr # 1000-100
COMPARE
100 10 Glass
$393.40CAD / Case of 48
Pyrex 1000-150 Brand 1000 Griffin Low-Form Beaker, 150 mL, 48/cs 3450226 Mfr # 1000-150
COMPARE
150 10 Glass
$358.03CAD / Case of 48
Pyrex 1000-250 Brand 1000 Griffin Low-Form Beaker, 250 mL, 48/cs 3450227 Mfr # 1000-250
COMPARE
250 25 Glass
$378.66CAD / Case of 48
Pyrex 1000-400 Brand 1000 Griffin Low-Form Beaker, 400 mL, 48/cs 3450228 Mfr # 1000-400
COMPARE
400 25 Glass
$443.49CAD / Case of 48
Pyrex 1000-600 Brand 1000 Griffin Low-Form Beaker, 600 mL, 36/cs 3450229 Mfr # 1000-600
COMPARE
600 50 Glass
$422.86CAD / Case of 36
Pyrex 1000-800 Brand 1000 Griffin Low-Form Beaker, 800 mL, 24/cs 3450230 Mfr # 1000-800
COMPARE
800 50 Glass
$388.97CAD / Case of 24
Showing 1 - 20 of 25