Pyrex 7079 Glass Serological Pipettes
Disposable/Reusable glass serological non-sterile, non-plugged pipettes
  • Pipettes do not have plugged-ends and come bulk-packed
  • Negative graduations for versatiity
  • Reference: ASTM E-714
These non-sterile pipettes are calibrated "to deliver" (TD) with blow out. Slender, fire-polished tips allow for fast and easy pipetting with burr-free, even flow rates.
NOTE: Pipetting by mouth can be hazardous to your health. We suggest using a mechanical pipetting device.
$719.01 - $1,115.36CAD / Case of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL)
Availability Pricing
Pyrex 7079-1N Disposable Glass Serological Pipette, 1 mL, Bulk, 1000/Cs 3456824 Mfr # 7079-1N
COMPARE
1
$962.12CAD / Case of 1000
Pyrex 7079-2N Disposable Glass Serological Pipette, 2 mL, Bulk, 700/Cs 3456825 Mfr # 7079-2N
COMPARE
2
$719.01CAD / Case of 700
Pyrex 7079-10N Disposable Glass Serological Pipette, 10 mL, Bulk, 720/Cs 3456827 Mfr # 7079-10N
COMPARE
10
$1,115.36CAD / Case of 720