PRO Scientific Universal Homogenizing Packages
Homogenize in microtubes, round-bottom tubes, or conical-bottom tubes
  • Great for processing all types of samples, from soft to tough
  • 316 stainless steel and PTFE probe construction allows sterilization with any method
Universal homogenizing packages come with Bio-Gen PRO200 homogenizers featuring a small yet powerful motor to process all types of samples, even tough and stringy. Use with various tube sizes from 1.5 to 50 mL. Homogenizers feature a separate ON/OFF switch allowing for the slide switch speed control to remain at your ideal homogenizing speed. Two saw-tooth generator probes (7 mm and 10 mm diameter, included) provide excellent cutting action during homogenizing, resulting in fast and complete sample breakdown. All probes are precision crafted from 316 SS and PTFE to be chemically compatible with your sample. Deluxe package includes a stand for hands-free operation.
$5,245.27 - $5,967.23CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
PRO Scientific Standard Universal Homogenizer Package, 1.5 to 15 mL Tubes; 120V 0475106 Mfr # PK-01200UHS
COMPARE
120 Standard Universal Homogenizer Package, 1.5 to 15 mL Tubes; 120V
$5,245.27CAD / Each
PRO Scientific Standard Universal Homogenizer Package, 1.5 to 15 mL Tubes; 220V 0475107 Mfr # PK-02200UHS
COMPARE
220 Standard Universal Homogenizer Package, 1.5 to 15 mL Tubes; 220V
$5,458.91CAD / Each
PRO Scientific Deluxe Universal Homogenizer Package, 1.5 to 15 mL Tubes; 120V 0475108 Mfr # PK-01200UHD
COMPARE
120 Deluxe Universal Homogenizer Package, 1.5 to 15 mL Tubes; 120V
$5,768.32CAD / Each
PRO Scientific Deluxe Universal Homogenizer Package, 1.5 to 15 mL Tubes; 220V 0475109 Mfr # PK-02200UHD
COMPARE
220 Deluxe Universal Homogenizer Package, 1.5 to 15 mL Tubes; 220V
$5,967.23CAD / Each