PRO Scientific PRO25D Digital Homogenizer
Precision and power, processing volume of 3.5L
  • Compact digital homogenizer unit with high watt motor
  • 0.05ml - 3.5L volume range
  • Bright LED display and user-friendly controls
  • Homogenize, emulsify, blend and/or mix
This post-mounted benchtop homogenizer features accurate digital control of speed for results you can count on. The powerful motor's torque can handle samples up to 50,000 mPas or 3.5 L maximum volume. Perfect for compounding, cell disruption, protein extraction, and pharmaceutical processing. 220V PRO250D Digital Homogenizer model is available. Max volume equivalent to H20/1 centipoise.
Choose a generator probe based on the sample volume and dispersion type (order separately). The standard quick-connect generator probes are precision crafted from 316 stainless steel and PTFE, ranging 5mm to 30mm in diameter. 2-year warranty on probe to homogenizer connection.
$5,488.38 - $5,893.56CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
PRO Scientific PRO25D Digital Homogenizer with Stand; 120V 0475130 Mfr # 01-25100
COMPARE
120
$5,488.38CAD / Each
PRO Scientific PRO25D Digital Homogenizer with Stand; 220V 0475135 Mfr # 01-25200
COMPARE
220
$5,893.56CAD / Each