Polyethylene Tape
Excellent for sealing or resealing jar lids. White tape stretches and conforms to wrap flasks, vacuum bottles, and other glass vessels. Use to reduce injury risk from implosion.
$299.83 - $464.12CAD / Carton of 12
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material Width (in)
Availability Pricing
COMPARE
108 Polyethylene 1
$299.83CAD / Carton of 12
COMPARE
108 Polyethylene 2
$464.12CAD / Carton of 12