PharmaFluor® Compression to NPT(M) Threaded Fittings, Straight Adapter, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
$59.67 - $103.51CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Flow Pattern Thread Size NPT(M) (Port 2) (InchShort)
Availability Pricing
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/2" OD x 1/2" NPT(M) 5014449 Mfr # G1077001
COMPARE
1/2 Straight 1/2
$74.77CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/2" OD x 1/4" NPT(M) 5014450 Mfr # G1077050
COMPARE
1/2 Straight 1/4
$77.35CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/2" OD x 3/4" NPT(M) 5014451 Mfr # G1077104
COMPARE
1/2 Straight 3/4
$76.98CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/2" OD x 3/8" NPT(M) 5014452 Mfr # G1077005
COMPARE
1/2 Straight 3/8
$74.77CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/4" OD x 1/2" NPT(M) 5014453 Mfr # G1077081
COMPARE
1/4 Straight 1/2
$70.35CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/4" OD x 1/4" NPT(M) 5014454 Mfr # G1077002
COMPARE
1/4 Straight 1/4
$60.41CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/4" OD x 1/8" NPT(M) 5014455 Mfr # G1077053
COMPARE
1/4 Straight 1/8
$60.41CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/4" OD x 3/8" NPT(M) 5014456 Mfr # G1077080
COMPARE
1/4 Straight 3/8
$64.09CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/8" OD x 1/4" NPT(M) 5014457 Mfr # G1077054
COMPARE
1/8 Straight 1/4
$59.67CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/8" OD x 1/8" NPT(M) 5014458 Mfr # G1077067
COMPARE
1/8 Straight 1/8
$59.67CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 3/8" OD x 1/2" NPT(M) 5014459 Mfr # G1077051
COMPARE
3/8 1/2
$72.93CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 3/8" OD x 1/4" NPT(M) 5014460 Mfr # G1077003
COMPARE
3/8 Straight 1/4
$64.83CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 3/4" OD x 3/4" NPT(M) 5014461 Mfr # G1077006
COMPARE
3/4 Straight 3/4
$103.51CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 3/4" OD x 1/2" NPT(M) 5014462 Mfr # G1077052
COMPARE
3/4 1/2
$98.72CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 3/8" OD x 3/8" NPT("M) 5014463 Mfr # G1077004
COMPARE
3/8 Straight 3/8
$67.78CAD / Each
Showing 1- 15 of 15