PharmaFluor® Compression to NPT(F) Threaded Fittings, Straight Adapter, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
$72.56 - $106.08CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Flow Pattern Thread size NPT(F) (port 2) (in)
Availability Pricing
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/2" OD x 1/2" NPT(F) 5014443 Mfr # G1077090
COMPARE
1/2 Straight 1/2
$102.77CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/2" OD x 1/4" NPT(F) 5014444 Mfr # G1077088
COMPARE
1/2 Straight 1/4
$106.08CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/4" OD x 1/2" NPT(F) 5014445 Mfr # G1077086
COMPARE
1/4 Straight 1/2
$83.25CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 1/4" OD x 1/4" NPT(F) 5014446 Mfr # G1077055
COMPARE
1/4 Straight 1/4
$72.56CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 3/8" OD x 1/4" NPT(F) 5014447 Mfr # G1077056
COMPARE
3/8 Straight 1/4
$83.25CAD / Each
PharmaFluor Compression to Threaded Fitting, Straight Adapter, PFA, 3/8" OD x 3/8" NPT(F) 5014448 Mfr # G1077057
COMPARE
3/8 Straight 3/8
$85.46CAD / Each