PharmaFluor® Compression Fittings, Straight Reducer, PFA
High-purity compression fittings designed for critical fluid and gas applications
  • Ultra-clean PFA material construction
  • Excellent chemical resistance
  • Tool-free installation for faster setups
  • Reliable performance in critical systems
PharmaFluor® PFA compression fittings are engineered to deliver reliable, contamination-free performance in critical fluid and gas handling systems. Manufactured from 100% virgin high-purity PFA, these fittings ensure exceptional chemical resistance and ultra-clean operation, making them ideal for applications in biopharmaceutical, semiconductor, chemical, and food processing environments. These fittings do not require any tools, enabling fast, efficient installation while maintaining secure, leak-resistant connections.
$89.88 - $120.82CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Compression Size / Tube Od (Port 2) (in) Flow Pattern
Availability Pricing
PharmaFluor Compression Fitting, Straight Reducer, PFA, 3/4" OD x 1/2" OD 5014438 Mfr # G1077103
COMPARE
1/2 3/4
$115.29CAD / Each
PharmaFluor Compression Fitting, Straight Reducer, PFA, 1/2" OD x 1/4" OD 5014439 Mfr # G1077062
COMPARE
1/4 1/2 Straight
$120.82CAD / Each
PharmaFluor Compression Fitting, Straight Reducer, PFA, 3/8" OD x 1/4" OD 5014440 Mfr # G1077061
COMPARE
1/4 3/8 Straight
$107.93CAD / Each
PharmaFluor Compression Fitting, Straight Reducer, PFA, 1/4" OD x 1/8" OD 5014441 Mfr # G1077060
COMPARE
1/8 1/4 Straight
$89.88CAD / Each
PharmaFluor Compression Fitting, Straight Reducer, PFA, 1/2" OD x 3/8" OD 5014442 Mfr # G1077063
COMPARE
3/8 1/2 Straight
$113.08CAD / Each