PFA In-Line Filter Holders
Nonwetting and nonsticking filter holder
  • Temperature range is –320 to 500°F (–196 to 260°C)
  • Withstands up to 65 psi
Suitable for organic solvents, acids, alkalies, or high-purity filtration. Change filter without disconnecting tubing with Tefzel® ETFE clamp nut. Prefiltering reservoir creates fluid turbulence to slow membrane clogging. PFA support grid snaps out easily for cleaning. Ferruled nuts accept 1/4'' OD PTFE or PFA tubing. Order filter membranes separately.
$558.93 - $688.07CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Filter Diameter (mm)
Availability Pricing
COMPARE
PFA 47
$558.93CAD / Each
COMPARE
PFA 50
$688.07CAD / Each