Orochem Reliasil™ C18 HPLC Columns
Rugged design with enhanced chemical stability and efficiency
  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for C18 phase
The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume. Choose from standard columns with a pore size of 90 Å or columns with a 300 Å pore size.
$1,012.22 - $9,577.04CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Orochem Reliasil™ HPLC Column, C18, 3 µm, 100 mm L x 3.0 mm ID, 90Å 0648643 Mfr # R3DG-101
COMPARE
3 100 3
$1,091.78CAD / Each
Orochem Reliasil™ HPLC Column, C18, 5 µm, 100 mm L x 3.0 mm ID, 90Å 0648644 Mfr # R5DG-121
COMPARE
3 100 5
$1,012.22CAD / Each
Orochem Reliasil™ HPLC Column, C18, 3 µm, 150 mm L x 4.6 mm ID, 90Å 0648645 Mfr # R3FI-101
COMPARE
4.6 150 3
$1,046.11CAD / Each
Orochem Reliasil™ HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID, 90Å 0648646 Mfr # R5FI-121
COMPARE
4.6 150 5
$1,046.11CAD / Each
Orochem Reliasil™ HPLC Column, C18, 5 µm, 250 mm L x 4.6 mm ID, 90Å 0648647 Mfr # R5HI-121
COMPARE
4.6 250 5
$1,196.39CAD / Each
Orochem Reliasil™ HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID, 300Å 0648648 Mfr # R5FI-126
COMPARE
4.6 150 5
$1,237.65CAD / Each
Orochem Reliasil™ HPLC Column, C18, 5 µm, 250 mm L x 4.6 mm ID, 300Å 0648649 Mfr # R5HI-126
COMPARE
4.6 250 5
$1,390.88CAD / Each
Orochem Reliasil™ HPLC Column, C18, 10 µm, 250 mm L x 10 mm ID, 90Å 0648650 Mfr # R10HL-147
COMPARE
10 250 10
$5,274.74CAD / Each
Orochem Reliasil™ HPLC Column, C18, 5 µm, 250 mm L x 10 mm ID, 90Å 0648651 Mfr # R9HL-147
COMPARE
10 250 5
$2,276.39CAD / Each
Orochem Reliasil™ HPLC Column, C18, 10 µm250 mm L x 21 mm ID, 90Å 0648652 Mfr # R10HT-147
COMPARE
21 250 10
$9,577.04CAD / Each
Orochem Reliasil™ HPLC Column, C18, 5 µm, 250 mm L x 21 mm ID, 90Å 0648653 Mfr # R9HM-147
COMPARE
21 250 5
$5,635.72CAD / Each
Showing 1- 11 of 11