Orochem OroSil™ C8 HPLC Columns
Compatible even with 100% aqueous mobile phase
  • Ideal for separation of polar, semi-polar and nonpolar compounds at low to medium pH
  • Specialized surface treatment provides high hydrophobicity
  • Suitable for C8 phase
These Orochem OroSil™ HPLC columns are uniquely designed for the separation of polar, semi-polar and nonpolar compounds at low to medium pH. With specialized surface treatment, the OroSil™ chemistry makes these columns compatible even with 100% aqueous mobile phase. Columns are known to be stable at pH 1.5-9.0.
$904.66 - $1,287.74CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Orochem OroSil™ HPLC Column, C8, 3 µm, 150 mm L x 4.6 mm ID 0648622 Mfr # O3FI-924
COMPARE
4.6 150 3
$1,043.16CAD / Each
Orochem OroSil™ HPLC Column, C8, 5 µm, 150 mm L x 4.6 mm ID 0648623 Mfr # O5FI-923
COMPARE
4.6 150 5
$1,105.04CAD / Each
Orochem OroSil™ HPLC Column, C8, 3 µm, 250 mm L x 4.6 mm ID 0648624 Mfr # O3HI-924
COMPARE
4.6 250 3
$1,287.74CAD / Each
Orochem OroSil™ HPLC Column, C8, 5 µm, 250 mm L x 4.6 mm ID 0648625 Mfr # O5HI-923
COMPARE
4.6 250 5
$1,224.39CAD / Each
Orochem OroSil™ HPLC Column, C8, 5 µm, 50 mm L x 3.0 mm ID 0648626 Mfr # O5BG-923
COMPARE
3 50 5
$904.66CAD / Each