Ohaus Scout® SPX and STX Portable Toploading Balances
Superior performance and easy operation
  • Removable high strength stainless steel weighing platform
  • Impact-resistant pan support
  • Integrated transportation/storage lock
  • Sealed front panel and molded spill ring help keep the Scout safe from spills
  • Designed to be stackable when utilizing the stacking and storage cover (optional on the SPX)
Designed for use in laboratory, industrial or education applications, the Scout balance is available in two models—the SPX with LCD and tactical buttons and the STX with an informative color touchscreen. All models feature transportation lock, stainless steel pan, menu and calibration lockout switch, slip-resistant and adjustable feet, leveling bubble, mechanical and software overload/underload protection, stability indicator, auto tare, low-battery indicator, auto shut-off, and user-selectable printing and communication settings. These balances feature multiple weighing modes including enhanced parts counting, check weighing, percent weighing, density determination, totalization, and display hold. With optional RS-232, USB host, USB device, Ethernet, and Bluetooth® connectivity kits.
$612.93 - $2,665.36CAD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Ohaus STX123  Scout Portable Balance 120 g x 0.001 g with Touchscreen 1101511 Mfr # STX123
COMPARE
120 115 / 230 0.001
$1,732.71CAD / Each
Ohaus STX223 Scout Portable Balance 220 g x 0.001 g with Touchscreen 1101512 Mfr # STX223
COMPARE
220 115 / 230 0.001
$1,941.93CAD / Each
Ohaus STX222 Scout Portable Balance 220 g x 0.01 g with Touchscreen 1101513 Mfr # STX222
COMPARE
220 115 / 230 0.01
$1,277.43CAD / Each
Ohaus STX422 Scout Portable Balance 420 g x 0.01 g with Touchscreen 1101514 Mfr # STX422
COMPARE
420 115 / 230 0.01
$1,784.28CAD / Each
Ohaus STX622 Scout Portable Balance 620 g x 0.01 g with Touchscreen 1101515 Mfr # STX622
COMPARE
620 115 / 230 0.01
$2,090.74CAD / Each
Ohaus STX1202 Scout Portable Balance 1200 g x 0.01 g with Touchscreen 1101516 Mfr # STX1202
COMPARE
1200 115 / 230 0.01
$2,388.37CAD / Each
Ohaus STX2202 ScoutPortable Balance 2200 g x 0.01 g with Touchscreen 1101517 Mfr # STX2202
COMPARE
2200 115 / 230 0.01
$2,665.36CAD / Each
Ohaus STX421 Scout Portable Balance 420 g x 0.1 g with Touchscreen 1101518 Mfr # STX421
COMPARE
420 115 / 230 0.1
$723.43CAD / Each
Ohaus STX621 Scout Portable Balance 620 g x 0.1 g with Touchscreen 1101519 Mfr # STX621
COMPARE
620 115 / 230 0.1
$1,081.47CAD / Each
Ohaus STX2201 Scout Portable Balance 2200 g x 0.1 g with Touchscreen 1101522 Mfr # STX2201
COMPARE
2200 115 / 230 0.1
$1,367.31CAD / Each
Ohaus STX6201 Scout Portable Balance 6200 g x 0.1 g with Touchscreen 1101523 Mfr # STX6201
COMPARE
6200 115 / 230 0.1
$2,161.46CAD / Each
Ohaus STX8200 Scout Portable Balance 8200 g x 1 g with Touchscreen 1101525 Mfr # STX8200
COMPARE
8200 115 / 230 1
$1,121.25CAD / Each
Ohaus SPX123 Scout Portable Balance 120 g x 0.001 g 1101527 Mfr # SPX123
COMPARE
120 115 / 230 0.001
$1,526.43CAD / Each
Ohaus SPX223 Scout Portable Balance 220 g x 0.001 g 1101528 Mfr # SPX223
COMPARE
220 115 / 230 0.001
$1,704.71CAD / Each
Ohaus SPX222 Scout Portable Balance 220 g x 0.01 g 1101529 Mfr # SPX222
COMPARE
220 115 / 230 0.01
$1,107.99CAD / Each
Ohaus SPX422 Scout Portable Balance 420 g x 0.01 g 1101532 Mfr # SPX422
COMPARE
420 115 / 230 0.01
$1,555.90CAD / Each
Ohaus SPX622 Scout Portable Balance 620 g x 0.01 g 1101533 Mfr # SPX622
COMPARE
620 115 / 230 0.01
$1,824.06CAD / Each
Ohaus SPX1202 Scout Portable Balance 1200 g x 0.01 g 1101534 Mfr # SPX1202
COMPARE
1200 115 / 230 0.01
$2,080.43CAD / Each
Ohaus SPX2202 Scout Portable Balance 2200 g x 0.01 g 1101535 Mfr # SPX2202
COMPARE
2200 115 / 230 0.01
$2,339.74CAD / Each
Ohaus SPX421 Scout Portable Balance 420 g x 0.1 g 1101536 Mfr # SPX421
COMPARE
420 115 / 230 0.1
$612.93CAD / Each
Showing 1 - 20 of 24