Ohaus Adventurer AX Analytical Balances
A modern weighing experience with multiple connectivity options and advanced application functions
  • GLP/GMP compliance via real-time clock and recording of sample, project names, and balance ID
  • Two USB ports (one on the front and one on the back) provide connectivity with both a PC and a portable external device at the same time
  • Cloning feature saves settings to USB flash drive for transfer to other Adventurer balances
  • Color touch screen offers easy and fast operation with or without gloves
  • Six mechanical keys provide tactile feedback and allow the operator to perform repetitive tasks
  • Illuminated level indicator located on the front panel makes adjustment easy in any lighting condition
  • Choose between internal and external calibration models
  • Advanced weighing functions include: part counting, percent, dynamic, density, check weighing, totalization, and formulation modes
  • Draftshield doors are extra wide providing unobstructed access to larger weighing vessels
  • Overall design allows for easy cleaning, minimizing contamination
Additional features include RS232 interface, integral weigh below hook, full housing in-use cover, removable stainless steel pan, die-cast metal bottom housing, security bracket, illuminated up-front level indicator, four adjustable feet, software lockout menus, stability indicator, software overload/underload indicators, user selectable environmental settings, audible indicator, user selectable brightness settings, auto dim, touch screen calibration, auto tare, user selectable operating language.
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus AX124 Adventurer Analytical Balance 120g x 0.1 Mg Autocal 1101900 Mfr # AX124
COMPARE
Internal 120 0.0001
$6,908.73CAD / Each
Ohaus AX124/E Adventurer Analytical Balance 120g x 0.1 Mg Excal 1101901 Mfr # AX124/E
COMPARE
External 120 0.0001
$5,700.55CAD / Each
Ohaus AX224N Adventurer Analytical Balance 220g x 0.1 Mg NTEP Autocal 1101902 Mfr # AX224N
COMPARE
Internal 220 0.0001
$8,091.86CAD / Each
Ohaus AX224 Adventurer Analytical Balance 220g x 0.1 Mg Autocal 1101903 Mfr # AX224
COMPARE
Internal 220 0.0001
$7,624.79CAD / Each
Ohaus AX224/E Adventurer Analytical Balance 220g x 0.1 Mg Excal 1101904 Mfr # AX224/E
COMPARE
External 220 0.0001
$6,258.96CAD / Each
Ohaus AX324 Adventurer Analytical Balance 320g x 0.1 Mg Autocal 1101905 Mfr # AX324
COMPARE
Internal 320 0.0001
$8,333.49CAD / Each