Nylon Spiral Brush
Nylon spiral brush cleans the screen and helps avoids clogs. Place brush on the screen cloth, and it automatically rotates due to the vibration. Note: Use with screen cloths finer than 60 mesh only.
$517.90CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) RPM Sieves Size Accepted (OD)
Availability Pricing
CE International Trading LS450N0028 Nylon Spiral Brush for Vibro-Separators, 60 mesh and finer 5997900 Mfr # LS450N0028
COMPARE
$517.90CAD / Each