North Sciences PRE Series High-Performance Lab/Pharmacy Refrigerators
PRE series high-performance pharmacy refrigerators with standard shelves are designed with features that support sample protection for storage requiring 2° to 8°C, according to CDC recommendations
  • Strong casters with brakes makes moving the refrigerators even easier Triple coated tempered glass with heat reflective film ensures the inner temperature remains stable and is anti-condensation Refrigeration block is fully replaceable Forced-air circulation with special air ducts guarantees inner temperature consistency Unique evaporator design results in better uniformity with no ice build-up
PRE series high-performance pharmacy refrigerators are designed with features that support sample protection for the storage of pharmaceuticals, vaccines, chemotherapy, and other medical and pharmacy-grade storage requiring 2° to 8°C, according to CDC recommendations. The controller allows the inner temperature to be adjusted by increments of 0.1° with multiple sensors to display and accurately control the temperature. A safety door lock prevents unauthorized access. Auto defrost function offers reliable performance in high ambient temperature and high humidity areas.
$7,216.66 - $13,720.21CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature - Refrigerator (° C) Power (VAC) Refrigerator Capacity (Liters)
Availability Pricing
North Sciences PRE Series High-Performance Lab/Pharmacy Refrigerator, 2°C to 10°C, 595 L, 110V RK-44261-61 Mfr # PRE-0500-NA
COMPARE
2 120 595
$7,216.66CAD / Each
North Sciences PRE Series High-Performance Lab/Pharmacy Refrigerator, 2°C to 10°C, 595 L, 220V RK-44261-62 Mfr # PRE-0500-EU
COMPARE
2 220 595
$7,216.66CAD / Each
North Sciences PRE Series High-Performance Lab/Pharmacy Refrigerator, 2°C to 10°C, 1100 L, 110V RK-44261-65 Mfr # PRE-1000-NA
COMPARE
2 120 1100
$13,720.21CAD / Each
North Sciences PRE Series High-Performance Lab/Pharmacy Refrigerator, 2°C to 10°C, 1100 L, 220V RK-44261-66 Mfr # PRE-1000-EU
COMPARE
2 220 1100
$13,720.21CAD / Each