Mettler Toledo MR Series Precision Toploading Balances
Delivers easy worry-free weighing with reliable results day after day
  • Large color touchscreen and intuitive interface simplify operation
  • Electromagnetic Force Compensation (EMFC) weighing cell provides accurate and reliable results
  • Fully automatic time- and temperature-controlled internal calibration adjustment ensures accuracy
  • Flexible integration options—connect using Ethernet, USB-A, USB-C, and RS-232
  • Sample ID and passcode protection produce GLP documentation of results
  • Leveling assistant feature indicates when the balance is not level and offers onscreen help
  • Overload protection guards the weighing cell against excess weight overload
  • Durable high-grade chemical-resistant die-cast aluminum housing
The MR series precision toploading balances are great for routine laboratory weighing. The bright 4.5” color TFT touchscreen with intuitive guidance enables you to operate quickly and efficiently, even when wearing gloves. An electromagnetic force compensation (EMFC) weighing cell with fully automated internal calibration technology (FACT) automatically adjusts the balance to compensate for a change in ambient temperature, to provide consistent reliable results.
Connect balance to a printer or computer using Ethernet, USB, or RS-232 connection for complete and error-free documentation to easily comply with GLP guidelines. Interfaces support a barcode reader, printer, PC, and other laboratory instruments for easy traceability and data handling. Nine embedded applications include essential weighing functions, statistics, and formulation. Restrict modification of the balance settings to authorized personnel only with the password protection feature.
A robust housing made of high-quality chemical-resistant aluminum protects the sensitive sensor from environmental influences and impacts. Smooth surfaces and rounded edges provide quick and easy cleaning. Built-in overload protection prolongs the life of your balance. Convenient power-saving mode minimizes energy usage and saves money.
$5,296.84 - $7,101.74CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Mettler Toledo MR203 Precision Toploading Balance, MR Series, 220g x 1mg with Touchscreen, Draft Shield, and FACT Internal Calibration 0262878 Mfr # 30666212
COMPARE
220 1 Precision Toploading Balance, 220 g x 1 mg with Draft Shield
$5,296.84CAD / Each
Mettler Toledo MR303 Precision Toploading Balance, MR Series, 320g x 1mg with Touchscreen, Draft Shield, and FACT Internal Calibration 0262879 Mfr # 30666215
COMPARE
320 1 Precision Toploading Balance, 320 g x 1 mg with Draft Shield
$5,768.32CAD / Each
Mettler Toledo MR503 Precision Toploading Balance, MR Series, 520g x 1mg with Touchscreen, Draft Shield, and FACT Internal Calibration 0262880 Mfr # 30666218
COMPARE
520 1 Precision Toploading Balance, 520 g x 1 mg with Draft Shield
$6,541.85CAD / Each
Mettler Toledo MR603 Precision Toploading Balance, MR Series, 620g x 1mg with Touchscreen, Draft Shield, and FACT Internal Calibration 0262881 Mfr # 30666221
COMPARE
620 1 Precision Toploading Balance, 620 g x 1 mg with Draft Shield
$6,799.69CAD / Each
Mettler Toledo MR1002 Precision Toploading Balance, MR Series, 1200g x 10mg with Touchscreen and FACT Internal Calibration 0262882 Mfr # 30666224
COMPARE
1200 10 Precision Toploading Balance, 1200 g x 10 mg
$5,429.44CAD / Each
Mettler Toledo MR2002 Precision Toploading Balance, MR Series, 2200g x 10mg with Touchscreen and FACT Internal Calibration 0262883 Mfr # 30666227
COMPARE
2200 10 Precision Toploading Balance, 2200 g x 10 mg
$5,930.39CAD / Each
Mettler Toledo MR3002 Precision Toploading Balance, MR Series, 3200g x 10mg with Touchscreen and FACT Internal Calibration 0262884 Mfr # 30666230
COMPARE
3200 10 Precision Toploading Balance, 3200 g x 10 mg
$6,210.34CAD / Each
Mettler Toledo MR4002 Precision Toploading Balance, MR Series, 4200g x 10mg with Touchscreen and FACT Internal Calibration 0262885 Mfr # 30666233
COMPARE
4200 10 Precision Toploading Balance, 4200 g x 10 mg
$6,453.45CAD / Each
Mettler Toledo MR6002 Precision Toploading Balance, MR Series, 6200g x 10mg with Touchscreen and FACT Internal Calibration 0262886 Mfr # 30666236
COMPARE
6200 10 Precision Toploading Balance, 6200 g x 10 mg
$7,101.74CAD / Each
Mettler Toledo MR6001 Precision Toploading Balance, MR Series, 6200g x 100mg with Touchscreen and FACT Internal Calibration 0262887 Mfr # 30666239
COMPARE
6200 100 Precision Toploading Balance, 6200 g x 100 mg
$5,318.94CAD / Each
Showing 1- 10 of 10