Mettler Toledo MA Series NTEP-Approved Standard Precision Toploading Balances
Simple and efficient balances for routine everyday weighing
  • NTEP-approved legal-for-trade balances meet government standards and requirements for commercial applications
  • Large, easy-to-read LCD hybrid touchscreen simplify operation
  • Electromagnetic Force Compensation (EMFC) weighing cell provides accurate and reliable results
  • Internal calibration ensure accuracy
  • Flexible integration options—connect using USB-A and RS-232
  • Automatically recognizes plugged in peripheral devices
  • Easily identify samples using the sample ID function
  • Overload protection guards the weighing cell against excess weight overload
  • High-quality materials and solid construction provide long service life
The MA series NTEP-approved precision toploading balances offer excellent repeatability, readability, and durability, making them perfect for routine tasks. The large, easy-to-read LCD hybrid touchscreen provides on-screen instructions for quick and efficient operation even when wearing gloves. An electromagnetic force compensation (EMFC) weighing cell and internal calibration provide consistent reliable results.
The USB-A and RS-232 interfaces allow easy transfer of data to a printer, computer, or USB storage device for complete and error-free documentation. Ten built-in applications include essential weighing functions, statistics, and formulation. Perform weighing operations below the balance with the included weighing hook. Password protection feature restricts modification of the balance settings to authorized personnel only.
Rugged housing, made of high-quality and durable materials, protects the sensitive sensor from environmental influences and impacts. Smooth surfaces and rounded edges provide quick and easy cleaning. Built-in overload protection prolongs the life of your balance. Convenient power-saving mode minimizes energy usage and saves money.
$3,359.33 - $13,569.92CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Mettler Toledo MA103/A Precision Toploading Balance, NTEP-Approved, MA Series, 120g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262941 Mfr # 30697421
COMPARE
120 1 Precision Toploading Balance, NTEP-Approved, 120 g x 1 mg with Draft Shield
$3,565.60CAD / Each
Mettler Toledo MA203/A Precision Toploading Balance, NTEP-Approved, MA Series, 220g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262942 Mfr # 30697424
COMPARE
220 1 Precision Toploading Balance, NTEP-Approved, 220 g x 1 mg with Draft Shield
$3,830.81CAD / Each
Mettler Toledo MA303/A Precision Toploading Balance, NTEP-Approved, MA Series, 320g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262943 Mfr # 30697427
COMPARE
320 1 Precision Toploading Balance, NTEP-Approved, 320 g x 1 mg with Draft Shield
$4,081.29CAD / Each
Mettler Toledo MA503/A Precision Toploading Balance, NTEP-Approved, MA Series, 520g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262944 Mfr # 30697430
COMPARE
520 1 Precision Toploading Balance, NTEP-Approved, 520 g x 1 mg with Draft Shield
$4,648.55CAD / Each
Mettler Toledo MA602/A Precision Toploading Balance, NTEP-Approved, MA Series, 620g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262945 Mfr # 30697441
COMPARE
620 10 Precision Toploading Balance, NTEP-Approved, 620 g x 10 mg
$3,359.33CAD / Each
Mettler Toledo MA1002/A Precision Toploading Balance, NTEP-Approved, MA Series, 1200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262946 Mfr # 30697444
COMPARE
1200 10 Precision Toploading Balance, NTEP-Approved, 1200 g x 10 mg
$3,845.55CAD / Each
Mettler Toledo MA2002/A Precision Toploading Balance, NTEP-Approved, MA Series, 2200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262947 Mfr # 30697447
COMPARE
2200 10 Precision Toploading Balance, NTEP-Approved, 2200 g x 10 mg
$4,258.10CAD / Each
Mettler Toledo MA3002/A Precision Toploading Balance, NTEP-Approved, MA Series, 3200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262948 Mfr # 30697450
COMPARE
3200 10 Precision Toploading Balance, NTEP-Approved, 3200 g x 10 mg
$4,361.23CAD / Each
Mettler Toledo MA4002/A Precision Toploading Balance, NTEP-Approved, MA Series, 4200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262949 Mfr # 30697453
COMPARE
4200 10 Precision Toploading Balance, NTEP-Approved, 4200 g x 10 mg
$4,611.71CAD / Each
Mettler Toledo MA6002/A Precision Toploading Balance, NTEP-Approved, MA Series, 6200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262950 Mfr # 30697456
COMPARE
6200 10 Precision Toploading Balance, NTEP-Approved, 6200 g x 10 mg
$5,031.63CAD / Each
Mettler Toledo MA5001/A Precision Toploading Balance, NTEP-Approved, MA Series, 5200g x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262951 Mfr # 30697459
COMPARE
5200 100 Precision Toploading Balance, NTEP-Approved, 5200 g x 100 mg
$3,683.48CAD / Each
Mettler Toledo MA12001L/A Precision Toploading Balance, NTEP-Approved, MA Series, 12.2kg x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262952 Mfr # 30697476
COMPARE
12200 100 Precision Toploading Balance, NTEP-Approved, 12.2 kg x 100 mg
$11,492.44CAD / Each
Mettler Toledo MA16001L/A Precision Toploading Balance, NTEP-Approved, MA Series, 16.2kg x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262953 Mfr # 30697479
COMPARE
16200 100 Precision Toploading Balance, NTEP-Approved, 16.2 kg x 100 mg
$12,509.08CAD / Each
Mettler Toledo MA32001L/A Precision Toploading Balance, NTEP-Approved, MA Series, 32.2kg x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262954 Mfr # 30697482
COMPARE
32200 100 Precision Toploading Balance, NTEP-Approved, 32.2 kg x 100 mg
$13,569.92CAD / Each
Mettler Toledo MA32000L/A Precision Toploading Balance, NTEP-Approved, MA Series, 32.2kg x 1g with LCD Hybrid Touchscreen and Internal Calibration 0262955 Mfr # 30697485
COMPARE
32200 1000 Precision Toploading Balance, NTEP-Approved, 32.2 kg x 1 g
$11,072.53CAD / Each
Showing 1- 15 of 15