Metrepak pH Buffer Capsules

No more wasted buffer
  • Make only the buffer you need—empty one capsule into 100 mL of water and stir
  • Accurate to ±0.02 pH
  • Ten capsules per vial
Prepare fresh buffer solutions with these handy pH buffer capsules. Empty the contents of one capsule into 100 mL of distilled water and stir. Buffer solutions are accurate to ±0.02 pH. Each vial contains ten capsules.
$35.14 - $39.34CAD / Vial of 10

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Metrepak 272,PH4 pH Calibration Buffer Capsules, 4.0, orange, 10/vial 0597712 Mfr # 272 PH4
COMPARE
pH Calibration Buffer Capsules, 4.0, orange, 10/vial
$38.68CAD / Vial of 10
Metrepak 273,PH7 pH Calibration Buffer Capsules, 7.0, green, 10/vial 0597714 Mfr # 273 PH7
COMPARE
pH Calibration Buffer Capsules, 7.0, green, 10/vial
$38.23CAD / Vial of 10
Metrepak 270-9.00 pH Calibration Buffer Capsules, 9.0, yellow, 10/vial 0597716 Mfr # 270-9.00
COMPARE
pH Calibration Buffer Capsules, 9.0, yellow, 10/vial
$35.14CAD / Vial of 10
Metrepak 274,PH10 pH Calibration Buffer Capsules, 10.0, blue, 10/vial 0597718 Mfr # 274 PH10
COMPARE
pH Calibration Buffer Capsules, 10.0, blue, 10/vial
$39.34CAD / Vial of 10
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat