Market Forge 16" Sterilmatic Benchtop Sterilizers
Fast preheat time—reaches 121°C (250°F) in less than 10 minutes
  • Heating elements never come in contact with water
  • ASME approved
These benchtop sterilizers are ideal for steam sterilization of wrapped or unwrapped dressings, labware, and liquids. Choose from the standard sterilizer with sterilizing temperature set at 121°C (250°F) or the laboratory sterilizer with an adjustable sterilizing temperature from 110 to 121°C (230 to 250°F).
Safety features include a door that locks until steam pressure is completely relieved from the chamber and low water-level cut-off. All models are rated for four-wire, three-phase operation. Sterilizer chambers are ASME-approved. Optional stainless steel stand, perforated tray, and basket sold separately.
$29,130.39 - $30,880.78CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Chamber Capacity (cu ft) Max Pressure (PSI) Power (VAC)
Availability Pricing
Market Forge 95-2678 Sterilmatic Benchtop Sterilizer, 16" Standard; 208V 1077500 Mfr # STM-E
COMPARE
3 15 208
$29,130.39CAD / Each
Market Forge 95-2902 Sterilmatic Benchtop Sterilizer, 16" Standard; 220/380V 1077505 Mfr # STM - EX
COMPARE
3 15 220 / 380
$30,880.78CAD / Each
Market Forge 95-3441 Sterilmatic Benchtop Sterilizer, 16" Adjustable; 240V 1077510 Mfr # STM-EL
COMPARE
3 8 240
$29,983.49CAD / Each