MadgeTech AVS140 Data Logging Autoclave Validation Systems
Provide data integrity, ensuring regulatory compliance
  • Stainless steel IP68-rated data loggers function in harsh environments
  • Software includes IQ/OQ/PQ protocols and tools for compliance with FDA 21 CFR 11
  • Software programmable immediate start or delay start up to 6 months
$3,433.00 - $9,348.66CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
MadgeTech AVS140-1 AVS140 Autoclave Validation System, 1 temperature data logger 1801102 Mfr # AVS140-1
COMPARE
500 ±0.18 (68 to 284), ±0.54 (-4 to 67.98), ±0.75 (-40 to 4.02) 0.02
$3,433.00CAD / Each
MadgeTech AVS140-6 AVS140 Autoclave Validation System, 5 temperature and 1 pressure 1801103 Mfr # AVS140-6
COMPARE
500 ±0.18 (68 to 284), ±0.54 (-4 to 67.98), ±0.75 (-40 to 4.02) 0.02
$9,348.66CAD / Each