Lascar Electronics Compact USB Data Loggers, Temperature - External Sensor
Direct-to-USB connection and easy-to-use functionality
  • User-programmable alarm thresholds
  • All data loggers come with software
Simply insert this compact, 5.3 in. (13.5 cm) long, logger into your computer's USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using the included, EasyLog, software.
$176.07 - $257.84CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Lascar Electronics EL-USB-TC Thermocouple Temperature USB Data Logger, 32,000+ readings, 6 month battery life 2303960 Mfr # EL-USB-TC
COMPARE
2462 ±2
$176.07CAD / Each
Lascar Electronics EL-USB-TC-LCD Thermocouple Data Logger with LCD Display, 32,000+ readings, 6 month battery life 2303964 Mfr # EL-USB-TC-LCD
COMPARE
2462 ±2 1
$257.84CAD / Each
Lascar Electronics EL-USB-TP-LCD Thermistor Data Logger with LCD Display 2303980 Mfr # EL-USB-TP-LCD
COMPARE
257 ±0.2 0.1
$238.32CAD / Each