Labconco Purifier HEPA-Filtered Enclosures
Protect you from biological hazards requiring Biosafety Level 1, 2, and 3 containment
  • EPA filter removes 99.99% of particulates down to 0.3 µm
Purifier enclosures protect you and your laboratory environment — they offer an economical alternative to Class II laminar flow cabinets when your applications do not require product protection. The filter indicator monitors the differential pressure across the filter and lets you know when to change the filter. Side panels and pivoting, angled sash are made of tempered safety glass; sash pivots upward 20" for easy cabinet loading. All units are shipped fully assembled. Order a spill-containing work surface and other accessories separately.
HEPA-Filtered Enclosures feature fluorescent lamp but do not include a UV light. They protect the user from hazardous chemical powders, dust, and allergens in diverse applications ranging from screening suspicious mail to weighing drug ingredients.
$11,290.59 - $14,025.20CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exhaust Volume (CFM) Hood Width (in) Power (VAC)
Availability Pricing
Labconco Purifier HEPA Filtered Enclosure, 2' W; 115 VAC 3350712 Mfr # 3980202
COMPARE
110 to 155 24 115
$11,290.59CAD / Each
Labconco Purifier HEPA Filtered Enclosure, 2' W, 230 VAC 3350714 Mfr # 3980222
COMPARE
110 to 155 24 230
$11,641.25CAD / Each
Labconco Purifier HEPA Filtered Enclosure, 3' W; 115 VAC 3350716 Mfr # 3980302
COMPARE
165 to 230 36 115
$12,634.32CAD / Each
Labconco Purifier HEPA Filtered Enclosure, 3' W, 230 VAC 3350718 Mfr # 3980322
COMPARE
165 to 230 36 230
$12,843.54CAD / Each
Labconco Purifier Purifer HEPA Filtered Enclosure, 4'W, 115V 3350720 Mfr # 3980402
COMPARE
220 to 310 48 115
$13,602.34CAD / Each
Labconco Purifier HEPA Filtered Enclosure, 4' W, 230 VAC 3350722 Mfr # 3980422
COMPARE
220 to 310 48 230
$14,025.20CAD / Each