Labconco Protector XStream Laboratory Hoods
Lab safety that saves energy and money
  • Lowest CFM requirement per corresponding face velocity (fpm) on the market
  • Low flow velocities mean less energy waste
Labconco Protector XStream Hoods are built for highest performance allowing them to operate at velocities as low as 60 fpm (minimum allowable face velocity per industry standards) while maintaining containment. Design negates the effects of air turbulence, vortexing, and interior air “roll.” Containment features include: Clean-Sweep™ sash track and sash handle that directs airflow into the hood; Eco-Foil™ air foil that corrects natural turbulence while keeping cords out of the way; and Opti-Zone™ baffle system with a unique slot pattern that balances face velocity so that less overall volume is required. The XStream passed strict containment testing, far exceeding industry standards.
Return on investment with the XStream comes not only through the savings realized by running a safe lab and purchasing durable equipment, but also through reduced operating expense. The true measure of fume hood energy consumption and operating expense is its total required volumetric rate (CFM). XStream’s low flow velocities mean less energy waste from hood air exhaust, but more importantly, it has the lowest CFM requirement per corresponding face velocity (fpm) on the market. Large energy savings are realized when comparing the XStream operating at 60 fpm to a more traditional hood at 100 fpm.
Hood features a large unobstructed sightline — 37.5” from work surface to header panel — and provides unobstructed viewing into the hood. Other features include dry powder-coated steel exterior, chemically resistant composition panel liner, pre-set rear downflow dual baffle, safety glass sash, pre-wired LED lighting rated to 50,000 hours, sash stop located at 18” sash opening position, and 12.81” ID exhaust connection(s).
$20,936.87 - $35,302.42CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Power (VAC)
Availability Pricing
Labconco Protector Xstream High-Performance Lab Hood, 4', Two Services and Duplex, 115V, 60Hz 0910020 Mfr # 110410002
COMPARE
59 48 115
$20,936.87CAD / Each
Labconco Protector Xstream High-Performance Lab Hood, 5', Two Services and Duplex, 115V, 60Hz 0910021 Mfr # 110510002
COMPARE
59 60 115
$23,176.42CAD / Each
Labconco Protector Xstream High-Performance Lab Hood, 6', Two Services and Duplex, 115V, 60Hz 0910022 Mfr # 110610002
COMPARE
59 72 115
$25,430.71CAD / Each
Labconco Protector Xstream High-Performance Lab Hood, 8', Two Services and Duplex, 115V, 60Hz 0910023 Mfr # 110810002
COMPARE
59 96 115
$35,302.42CAD / Each