Labconco Protector Downdraft Powder Stations
Fingerprint powdering of large items is no longer a challenge!
  • Safely work with nuisance powders
  • Open-side, open-top design allows unrestricted work space and accommodates oversized items
The HEPA filtered, open design of the downdraft powder stations safely contain powders while giving the freedom of an open work area. With the addition of the optional transition connector 28616-38, multiple downdraft powder stations can be joined together to create a longer, continuous work station.
Durable workstations are constructed of epoxy coated steel and include a removable stainless steel work surface. Beneath the work surface is a powder collection tray that is removable for easy clean up. The 75-watt blower is adjustable to optimize the life of each HEPA filter. A filter alarm light alerts you to low airflow conditions prompting a filter change. Each downdraft powder station comes complete with a prefilter and a HEPA filter; available in 2-ft (0.6-m) and 3-ft (0.9-m) widths
Note This product is for nuisance powders only. It is not to be used for biohazards, toxic chemicals or radioisotopes.
$7,190.14CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Description
Availability Pricing
Labconco Protector Downdraft Powder Station, 3-ft Wide, 115 V 2861606 Mfr # 3410001
COMPARE
13.3125 36 Downdraft Powder Station, 3-ft Wide, 115 V
$7,190.14CAD / Each