Labconco Precise HEPA-Filtered Glove Boxes
Laboratory protection at an economical price
  • Customizable units for specific applications
  • Easily removable window for equipment loading
Glove boxes feature a liner of one-piece molded, medium density polyethylene and a dry powder epoxy-coated steel exterior. The chamber features a 1/4" thick laminated safety glass viewing window with powder-coated steel frame and neoprene gasket. The cutout is 26 2/5"W x 12 1/2"H, and is easily removable for loading equipment. Space-saving inner and outer transfer chamber (11" nominal ID x 12"L) doors pivot upward, and are counterbalanced and equipped with quick-latches.
Units also feature an interior ceiling mounted electrical duplex receptacle with a 5 amp maximum load, and a flourescent lamp that is easily replaceable from the top of the unit. The black, chemical resistant work surface is sealed to the glove box floor, allowing for easy cleaning.
$17,570.18CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Width (in) Power (VAC)
Availability Pricing
Labconco 5220200 Precise HEPA-Filtered Glove Box, 33.5" Wide; 115 VAC 3476350 Mfr # 5220200
COMPARE
33.5 115
$17,570.18CAD / Each
Labconco 5220220 Precise HEPA-Filtered Glove Box, 33.5" Wide; 230 VAC 3476360 Mfr # 5220220
COMPARE
33.5 230
$17,570.18CAD / Each