Labconco Filters for Purifier® Clean Benches

Ensure clean airflow with filters and prefilters
  • Fits in clean benches to help protect critical, non-hazardous samples
$50.10 - $3,514.04CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Type Compatible With Width (InchShort)
Availability Pricing
Labconco 3718000 Replacement Prefilter for 24" Purifier® Vertical Clean Benches 0911464 Mfr # 3718000
COMPARE
Prefilter 24" Purifier® vertical clean benches 24
$96.36CAD / Each
Labconco Replacement Prefilter for 36" Purifier® Vertical Clean Benches 0911466 Mfr # 3935701
COMPARE
Prefilter 36" Purifier® vertical clean benches 36
$66.74CAD / Each
Labconco Replacement Prefilter for 36" PuriCare® Open Access Stations and Purifier® Horizontal Clean Benches 0911490 Mfr # 3768900
COMPARE
Prefilter 36" PuriCare® open access stations and Purifier® horizontal clean benches 36
$50.10CAD / Each
Labconco Purifier® Replacement Prefilter for 48" or 72" Purifier® Horizontal Clean Benches 0911491 Mfr # 3768901
COMPARE
Prefilter 48" or 72" Purifier® horizontal clean benches 72
$53.48CAD / Each
Labconco Purifier® HEPA Filter for 72" Purifier® Horizontal Clean Benches 0911686 Mfr # 1489004
COMPARE
HEPA filter 72" Purifier® horizontal clean benches 72
$3,514.04CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat