Labconco Filtered Balance Systems
Minimize risk of exposure to toxic powders and fumes while maintaining accurate balance readings
  • Static-dissipating design reduces weighing errors
These space-saving, self-contained balance systems feature an air foil to keep you safe while working with hazardous materials. The built-in blower and HEPA filter save space, making them perfect for tight areas or locations where ductwork is not available. The HEPA filter removes 99.99% of particles 0.3-micron or larger. Negative pressure design ensures that if a leak develops, unfiltered air is filtered before release. Unique bag-in/bag-out filter disposal system provides safe filter replacement without exposing you to contaminants. Cabinet design minimizes electrostatic effects and vibration to enable precise weighing results.
Enclosures are constructed of an epoxy-coated aluminum frame, and steel rear plenum and baffle. Top-mounted blower with speed control and HEPA filter reduce space requirements, no separate filtered exhauster or outside ventilation needed. Sash and sides are made of 1/4"-thick tempered safety glass, providing better resistance to fire and chemicals than acrylic. Units with 30" interior depth include a mounted waste chute. Two utility ports are included for cords, cables, and tubing.
Models with deep 23-13/32" interior depth provide easy loading of analytical balances; the deeper 30” models handle larger equipment. Ergonomic, angled sash pivots upward offering a 19" loading height on the 2-, 3-, and 4-ft models, while the 5- and 6-foot models have a gas-assisted lift and loading height of 27-45/64".
Choose enclosures with an optional airflow monitor to alert you of face velocities outside of a preset range. Models with no monitor include a front-mounted pressure gauge, which indicates loading of the HEPA filter. The Guardian airflow monitor provides continuous airflow monitoring of airflow with a LED monitor that stays green within the set range; LED changes to red and an audible alarm sounds if airflow is out of range. The Guardian 1000 Digital airflow monitor features a digital display of the airflow at face velocity and a RS-232 interface for data transmission to your computer; the monitor provides an audible and visual alarm if the airflow is outside the preset range.
To complete your Filtered Balance System, choose a base stand or cabinet, filters, and work surfaces from Labconco Filtered Balance Systems Accessories. These are required to properly use your balance system.
$11,536.64 - $38,772.26CAD / Each
42 variations of this product are available.
See All Product Options Below
42 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Height (in) Exterior Width (in) Description
Availability Pricing
Labconco Xpert 2-ft Filtered Balance System, 115V 60hz 0910200 Mfr # 3950200
COMPARE
37.125 24 2-ft Filtered Balance System, 115 V, 60 Hz
$11,536.64CAD / Each
Labconco Xpert 2-ft Filtered Balance System With Guardian Airflow Monitor, 115V 60hz 0910210 Mfr # 3950201
COMPARE
37.125 24 2-ft Filtered Balance System With Guardian Airflow Monitor, 115 V, 60 Hz
$13,297.34CAD / Each
Labconco Xpert 2-ft Filtered Balance System With Digital Airflow Monitor, 115V 60hz 0910220 Mfr # 3950202
COMPARE
37.125 24 2-ft Filtered Balance System With Digital Airflow Monitor, 115 V, 60 Hz
$15,905.25CAD / Each
Labconco Xpert 2-ft Filtered Balance System, 230V 50hz 0910230 Mfr # 3950220
COMPARE
37.125 24 2-ft Filtered Balance System, 230 V, 50 Hz
$11,949.19CAD / Each
Labconco Xpert 2-ft Filtered Balance System With Guardian Airflow Monitor, 230V 50hz 0910240 Mfr # 3950221
COMPARE
37.125 24 2-ft Filtered Balance System With Guardian Airflow Monitor, 230 V, 50 Hz
$13,717.26CAD / Each
Labconco Xpert 2-ft Filtered Balance System With Digital Airflow Monitor, 230V 50hz 0910250 Mfr # 3950222
COMPARE
37.125 24 2-ft Filtered Balance System With Digital Airflow Monitor, 230 V, 50 Hz
$16,347.26CAD / Each
Labconco Xpert 3-ft Filtered Balance System, 115V 60hz 0910300 Mfr # 3950300
COMPARE
37.125 36 3-ft Filtered Balance System, 115 V, 60 Hz
$12,803.76CAD / Each
Labconco Xpert 3-ft Filtered Balance System With Guardian Airflow Monitor, 115V 60hz 0910310 Mfr # 3950301
COMPARE
37.125 36 3-ft Filtered Balance System With Guardian Airflow Monitor, 115 V, 60 Hz
$14,586.56CAD / Each
Labconco Xpert 3-ft Filtered Balance System With Digital Airflow Monitor, 115V 60hz 0910320 Mfr # 3950302
COMPARE
37.125 36 3-ft Filtered Balance System With Digital Airflow Monitor, 115 V, 60 Hz
$17,209.20CAD / Each
Labconco Xpert 3-ft Filtered Balance System, 230V 50hz 0910330 Mfr # 3950320
COMPARE
37.125 36 3-ft Filtered Balance System, 230 V, 50 Hz
$13,223.68CAD / Each
Labconco Xpert 3-ft Filtered Balance System With Guardian Airflow Monitor, 230V 50hz 0910340 Mfr # 3950321
COMPARE
37.125 36 3-ft Filtered Balance System With Guardian Airflow Monitor, 230 V, 50 Hz
$15,013.84CAD / Each
Labconco Xpert 3-ft Filtered Balance System With Digital Airflow Monitor, 230V 50hz 0910350 Mfr # 3950322
COMPARE
37.125 36 3-ft Filtered Balance System With Digital Airflow Monitor, 230 V, 50 Hz
$17,651.21CAD / Each
Labconco Xpert 4-ft Filtered Balance System, 115V 60hz 0910400 Mfr # 3950400
COMPARE
37.125 48 4-ft Filtered Balance System, 115 V, 60 Hz
$14,070.87CAD / Each
Labconco Xpert 4-ft Filtered Balance System With Guardian Airflow Monitor, 115V 60hz 0910410 Mfr # 3950401
COMPARE
37.125 48 4-ft Filtered Balance System With Guardian Airflow Monitor, 115 V, 60 Hz
$15,861.04CAD / Each
Labconco Xpert 4-ft Filtered Balance System With Digital Airflow Monitor, 115V 60hz 0910420 Mfr # 3950402
COMPARE
37.125 48 4-ft Filtered Balance System With Digital Airflow Monitor, 115 V, 60 Hz
$18,476.31CAD / Each
Labconco Xpert 4-ft Filtered Balance System, 230V 50hz 0910431 Mfr # 3950420
COMPARE
37.125 48 4-ft Filtered Balance System, 230 V, 50 Hz
$14,527.63CAD / Each
Labconco Xpert 4-ft Filtered Balance System With Guardian Airflow Monitor, 230V 50hz 0910440 Mfr # 3950421
COMPARE
37.125 48 4-ft Filtered Balance System With Guardian Airflow Monitor, 230 V, 50 Hz
$16,288.33CAD / Each
Labconco Xpert 4-ft Filtered Balance System With Digital Airflow Monitor, 230V 50hz 0910450 Mfr # 3950422
COMPARE
37.125 48 4-ft Filtered Balance System With Digital Airflow Monitor, 230 V, 50 Hz
$18,910.96CAD / Each
Labconco Xpert Filtered Balance System, 5-ft L x 23" D; 115V 60hz 0910500 Mfr # 3950500
COMPARE
46 60 Filtered Balance System, 5-ft L x 23" D; 115 V, 60 Hz
$25,003.43CAD / Each
Labconco Xpert Filtered Balance System With Airflow Monitor, 5-ft L x 30" D; 115V 60hz 0910510 Mfr # 3950501
COMPARE
46 60 Filtered Balance System With Airflow Monitor, 5-ft L x 30" D; 115 V, 60 Hz
$26,241.08CAD / Each
Showing 1 - 20 of 42