Labconco Double HEPA Filtration Kits for XPert® and RXPert™ Enclosures
Provides a second level of HEPA filtration for added safety
  • For filed installation atop XPert filtered balance systems and XPert bulk powder filtered systems
  • Meet USP <800> standard for redundant HEPA filter in series
Kit adds 12.9" to height to existing system.
$7,057.54 - $9,407.60CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Compatible With Width (InchShort)
Availability Pricing
Labconco Double HEPA Filtration Kit for 24" XPert® and RXPert™ Enclosures 3252195 Mfr # 3966102
COMPARE
24" XPert® filtered balance systems, Xpert bulk powder filtered systems, and RXPert™ enclosures 24
$7,057.54CAD / Each
Labconco Double HEPA Filtration Kit for 36" XPert® and RXPert™ Enclosures 3252196 Mfr # 3966103
COMPARE
36" XPert® filtered balance systems, Xpert bulk powder filtered systems, and RXPert™ enclosures 36
$7,654.26CAD / Each
Labconco Double HEPA Filtration Kit for 48" XPert® and RXPert™ Enclosures 3252197 Mfr # 3966104
COMPARE
48" XPert® filtered balance systems, Xpert bulk powder filtered systems, and RXPert™ enclosures 48
$8,228.88CAD / Each
Labconco Double HEPA Filtration Kit for 60" XPert® and RXPert™ Enclosures 3252198 Mfr # 3966105
COMPARE
60" XPert® filtered balance systems, Xpert bulk powder filtered systems, and RXPert™ enclosures 60
$8,825.61CAD / Each
Labconco Double HEPA Filtration Kit for 72" XPert® and RXPert™ Enclosures 3252199 Mfr # 3966106
COMPARE
72" XPert® filtered balance systems, Xpert bulk powder filtered systems, and RXPert™ enclosures 72
$9,407.60CAD / Each