Kimax Amber Dropping Bottles
Amber borosilicate glass bottles protect light sensitive chemicals. They are supplied with a 20-400 GPI screw cap, rubber nipple, and 1-mL dropper made of glass. Bottles meet ASTM E 438, Type II and USP Type III standards.
ITEM(S) ON SALE
$74.00 - $534.84CAD / Case of 72
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
DWK Life Sciences (Kimble) 15040P-30 30 mL Amber Glass Dropping Bottle, Plastic dropper 3452348 Mfr # 15040P-30
COMPARE
30 Glass 30 mL Amber Glass Dropping Bottle, Plastic dropper
$534.84CAD / Case of 72
DWK Life Sciences (Kimble) 15040P-60 60 mL Amber Glass Dropping Bottle, Plastic dropper 3452350 Mfr # 15040P-60
COMPARE
60 Glass 60 mL Amber Glass Dropping Bottle, Plastic dropper
$74.00CAD / Case of 12
$93.27CAD / Case of 12