Katanax® X-300 Multiposition Electric Fusion Fluxers
Prepare up to three disks simultaneously
  • Fuse samples and mix solutions without flames
  • Fast initial heat-up time of under 15 minutes
  • Ceramic mold holders eliminate metal contamination
  • Low noise level during heating, melting and standby
Easily prepare fused beads for X-ray fluorescence (XRF) analysis or solutions for AA, ICP, and wet chemistry analysis with the multiposition fusion fluxers. These versatile fluxers come prewired for three positions to meet your expanding laboratory needs. Dynamic Temperature Profile (DTP) control and automatic power calibration help maintain excellent temperature uniformity.
Built with user safety in mind, fluxers do not use propane or LPG so there are no pipes and no risk of gas leakage or open flames. An integrated auto-locking safety shield with viewport offers added protection. Extraction chimneys allow for direct ventilation of halogens from each crucible position. A tiltable control panel and a color LCD touch-screen interface with icons and menus make it simple to operate. Select from five languages and use preloaded or customized fusion methods. Constant display of temperature makes it easy to monitor the progress. Easily update firmware via USB connection.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Description
Availability Pricing
Environmental Express Katanax® Two-Position Electric Fusion Fluxer, Two Mold Holders 0457881 Mfr # X-300D
COMPARE
1200 Multiposition Electric Fusion Fluxer, Two Disk Molds
Call For Price
Environmental Express Katanax® Two-Position Electric Fusion Fluxer, Two Beakers 0457882 Mfr # X-300D-S
COMPARE
1200 Multiposition Electric Fusion Fluxer, Two Beakers
Call For Price
Environmental Express Katanax® One-Position Electric Fusion Fluxer, One Mold Holder 0457883 Mfr # X-300M
COMPARE
1200 Multiposition Electric Fusion Fluxer, One Disk Mold
Call For Price
Environmental Express Katanax® One-Position Electric Fusion Fluxer, One Beaker 0457884 Mfr # X-300M-S
COMPARE
1200 Multiposition Electric Fusion Fluxer, One Beaker
Call For Price
Environmental Express Katanax® Three-Position Electric Fusion Fluxer, Three Mold Holders 0457885 Mfr # X-300T
COMPARE
1200 Multiposition Electric Fusion Fluxer, Three Disk Molds
Call For Price
Environmental Express Katanax® Three-Position Electric Fusion Fluxer, Three Beakers 0457886 Mfr # X-300T-S
COMPARE
1200 Multiposition Electric Fusion Fluxer, Three Beakers