Katanax® Platinum Molds for Katanax® Fluxers

Use molds during borate fusion to form glass disks for XRF analysis
  • Made of 95% Pt-5% Au—the standard nonwetting alloy for borate fusions
Molds have 6 mm depth and have outer diameter ranging from 39 to 49 mm. Select heavy models with thicker walls and bottom for greater durability, longer lifespan, and better temperature control.
$2,322.06 - $4,082.76CAD / Each

13 variations of this product are available.

See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Inside Rim Diameter (mm) Description
Availability Pricing
COMPARE
Mold, 30mm, 95%Pt-5%Au, shallow (18g)
$2,322.06CAD / Each
COMPARE
Mold, 32mm, 95%Pt-5%Au, shallow (21g)
$2,652.10CAD / Each
COMPARE
Mold, 35mm, 95%Pt-5%Au, shallow (25g)
$3,092.65CAD / Each
COMPARE
Mold, 40mm, 95%Pt-5%Au, shallow (34g)
$4,082.76CAD / Each
COMPARE
Mold, 20mm, 95%Pt-5%Au, shallow (34g)
$4,082.76CAD / Each
Katanax Mold, 95% Pt-5% Au, 30 mm, Regular, 18 g 0457839 Mfr # KP1003A
COMPARE
30 Mold, 95% Pt-5% Au, 30 mm, Regular, 18 g
Call For Price
Katanax Mold, 95% Pt-5% Au, 30 mm, Heavy, 30 g 0457840 Mfr # KP1004A
COMPARE
30 Mold, 95% Pt-5% Au, 30 mm, Heavy, 30 g
Call For Price
Katanax Mold, 95% Pt-5% Au, 32 mm, Regular, 21 g 0457841 Mfr # KP1005A
COMPARE
32 Mold, 95% Pt-5% Au, 32 mm, Regular, 21 g
Call For Price
Katanax Mold, 95% Pt-5% Au, 32 mm, Heavy, 36 g 0457842 Mfr # KP1006A
COMPARE
32 Mold, 95% Pt-5% Au, 32 mm, Heavy, 36 g
Call For Price
Katanax Mold, 95% Pt-5% Au, 35 mm, Regular, 25 g 0457843 Mfr # KP1007A
COMPARE
35 Mold, 95% Pt-5% Au, 35 mm, Regular, 25 g
Call For Price
Katanax Mold, 95% Pt-5% Au, 35 mm, Heavy, 40 g 0457844 Mfr # KP1008A
COMPARE
35 Mold, 95% Pt-5% Au, 35 mm, Heavy, 40 g
Call For Price
Katanax Mold, 95% Pt-5% Au, 40 mm, Regular, 34 g 0457845 Mfr # KP1009A
COMPARE
40 Mold, 95% Pt-5% Au, 40 mm, Regular, 34 g
Call For Price
Katanax Mold, 95% Pt-5% Au, 40 mm, Heavy, 48 g 0457846 Mfr # KP1010A
COMPARE
40 Mold, 95% Pt-5% Au, 40 mm, Heavy, 48 g
Call For Price
Showing 1- 13 of 13

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat