Julabo SW-series Reciprocal Shaker Water Baths
Removable shaking insert can be easily prepared outside the bath
  • Adjustable temperature, shaking frequency, and countdown timer
  • One point temperature calibration for measurement accuracy
  • Shaking stroke at 15 mm with 20 to 200 rpm frequency
  • Low water-level warning and cut-off functions
These shaking baths were designed with features that make everyday laboratory task easier; from the durable side handles that allow the bath to be easily moved, to the removable shaking insert for easy setup, and the easy access drain.
Easy to use PID temperature controller with an LED display and the option of a one point temperature calibration ensure maximum accuracy. RS232 interface available for temperature data transfer.
Order bath cover and other accessories separately.
$9,395.81 - $11,759.13CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
Julabo SW22 Shaking Water Bath, ±0.2 C, 115V 1214900 Mfr # 9550322.2
COMPARE
115 Shaking Water Bath, ±0.2 C, 115V
$9,395.81CAD / Each
Julabo SW23 Shaking Water Bath, ±0.02C, 115V 1214910 Mfr # 9550323.2
COMPARE
115 Shaking Water Bath, ±0.02C, 115V
$11,759.13CAD / Each
Julabo SW23 Shaking Water Bath, ±0.02C, 230V 1214915 Mfr # 9550323.3
COMPARE
230 Shaking Water Bath, ±0.02C, 230V
$11,759.13CAD / Each