Julabo MAGIO™ MS / MX Heating Circulators
Extremely powerful pressure/suction pump perfect for external heating
  • Performance values up to 31 L/min or 0.92 bar (pressure)/-0.4 bar (suction)
  • All wetted parts are constructed of stainless steel for chemical resistance
  • High-resolution, TFT touch-screen display features a multi-lingual user interface
  • Remote control and data management made easy with USB interface, RS-232, and Ethernet
  • Pt100 external sensor connection for measurement and control
The MAGIO™ heating circulators are characterized by premium quality and high performance combined with intuitive usability. The units offer extra powerful pressure and suction pumps to meet the most challenging demands for temperature control of external applications. Samples can also be temperature controlled in the insulated and closed bath tank.
$11,958.03 - $14,347.87CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L)
Availability Pricing
Julabo MS-BC4 Heating Circulator with 4 L Bath Tank and Lid; with 1 kW Heating Power to +300 °C RK-12152-83 Mfr # 9032504.22
COMPARE
100 to 115 4.5
$11,958.03CAD / Each
Julabo MS-BC4 Heating Circulator with 4 L Bath Tank and Lid; with 2 kW Heating Power to +300 °C RK-12152-84 Mfr # 9032504.33
COMPARE
200 / 230 4.5
$11,958.03CAD / Each
Julabo MX-BC6 Heating Circulator with 6 L Bath Tank and Lid; with 3 kW Heating Power to +300 °C RK-12152-85 Mfr # 9033506.33
COMPARE
200 / 230 6
$12,733.04CAD / Each
Julabo MX-BC12 Heating Circulator with 12 L Bath Tank and Lid; with 3 kW Heating Power to +300 °C RK-12152-86 Mfr # 9033512.33
COMPARE
200 / 230 12
$13,631.80CAD / Each
Julabo MX-BC26 Heating Circulator with 26 L Bath Tank and Lid; with 3 kW Heating Power to +300 °C RK-12152-87 Mfr # 9033526.33
COMPARE
200 / 230 26
$14,347.87CAD / Each