Julabo CORIO™ CP-BC Heating Circulators
Easy pump change-over from internal to external circulation
  • Easily view bright white LED display from across the room
  • Remote control and data management made easy with USB and RS-232 interfaces
  • Meets Class III (FL) according to DIN 12876-1
The CORIO™ CP-BC heating circulators can easily be switched between internal and external circulation. All operating controls and safety functions can be easily accessed from the front of unit. One power switch controls the circulator and refrigeration unit while the pump control lever, located below the bright digital LED display, makes changing between internal and external circulation simple. Inclined pump connections (M16x1) facilitate the connection of external applications.
Other features include a front-mounted drain tap for removing bath fluids without the use of tools, USB and RS-232 interfaces for remote control and data management, removable front magnetic grid for easy cleaning and maintenance, lockable power plug, and an early warning system for low liquid level.
$8,146.37 - $10,575.99CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L) Description
Availability Pricing
Julabo CORIO™ CP-BC4 Heating Circulator, 4.5 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz 1215401 Mfr # 9013504.22
COMPARE
115 4.5 Heating Circulator, 4.5 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz
$8,146.37CAD / Each
Julabo CORIO™ CP-BC4 Heating Circulator, 4.5 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz 1215402 Mfr # 9013504.33
COMPARE
230 4.5 Heating Circulator, 4.5 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz
$8,146.37CAD / Each
Julabo CORIO™ CP-BC6 Heating Circulator, 6 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz 1215403 Mfr # 9013506.22
COMPARE
115 6 Heating Circulator, 6 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz
$8,781.40CAD / Each
Julabo CORIO™ CP-BC6 Heating Circulator, 6 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz 1215404 Mfr # 9013506.33
COMPARE
230 6 Heating Circulator, 6 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz
$8,781.40CAD / Each
Julabo CORIO™ CP-BC12 Heating Circulator, 12 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz 1215405 Mfr # 9013512.22
COMPARE
115 12 Heating Circulator, 12 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz
$9,506.31CAD / Each
Julabo CORIO™ CP-BC12 Heating Circulator, 12 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz 1215406 Mfr # 9013512.33
COMPARE
230 12 Heating Circulator, 12 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz
$9,506.31CAD / Each
Julabo CORIO™ CP-BC26 Heating Circulator, 26 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz 1215407 Mfr # 9013526.22
COMPARE
115 26 Heating Circulator, 26 L Tank, 20 to 200 °C, 1 kW Heater; 100-115 VAC, 50/60 Hz
$10,575.99CAD / Each
Julabo CORIO™ CP-BC26 Heating Circulator, 26 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz 1215408 Mfr # 9013526.33
COMPARE
230 26 Heating Circulator, 26 L Tank, 20 to 200 °C, 2 kW Heater; 200-230 VAC, 50/60 Hz
$10,575.99CAD / Each
Showing 1- 8 of 8