Inlet Foreline Cold Traps for Vacuum Pumps
  • Protect vacuum pump against damages from contaminants
These traps are easy to clean—remove from the vacuum pump and clean at the end of each day to prevent trapped oil vapors from damaging the pump. Dry ice traps have a removable 3-qt well for dry ice/alcohol slurry. Liquid nitrogen cold traps efficiently trap acidic vapors and lower the vacuum pump base pressure.
$2,438.46CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Material Power (VAC) Description
Availability Pricing
Welch 1420H-14 Foreline dry ice cold trap, 1/2" OD straight tube connection 7920930 Mfr # 1420H-14
COMPARE
Foreline dry ice cold trap, 1/2" OD straight tube connection
$2,438.46CAD / Each