Imada Cap Torque Tester
FEATURES
  • Programmable high/low setpoints with LED indicators for quick GO/NO GO testing
  • 1000 Data memory for recall and download
  • Accuracy: ±0.5% F.S. ±1 LSD
  • Clamps containers 1/2" to 8" in diameter
  • lbf-in, kgf-cm, and N-cm (selectable)
  • RS-232, Digimatic and analog outputs
  • Clockwise and counterclockwise operation
  • Real time and peak mode (selectable)
  • Tare function
  • AC adapter/charger and carrying case
$2,652.10CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-cm)
Availability Pricing
Imada DTXS-15 Digital Cap Torque Tester, 15 lbf-in 5984830 Mfr # DTXS-15
COMPARE
15 200
$2,652.10CAD / Each
Imada DTXS-40 Digital Cap Torque Tester, 40 lbf-in 5984831 Mfr # DTXS-40
COMPARE
40 500
$2,652.10CAD / Each
Imada DTXS-85 Digital Cap Torque Tester, 85 lbf-in 5984832 Mfr # DTXS-85
COMPARE
85 1000
$2,652.10CAD / Each