IKA® CBC 5 Series Refrigerated/Heating Circulating Baths
Powerful cooling/heating circulator for external applications requiring temperatures from –25 to 200°C
  • Variable-speed high performance pressure/suction pump
  • Large display for easy setting and viewing of parameters
  • RS-232 and USB interfaces—connect to a PC to control the recirculating chillers
  • Feed hopper and bleed valve offer safe and clean handling of the thermofluid
  • Easy access air filter makes cleaning convenient
  • Safety features include low liquid level indicator and safety temperature set
Use CRC 5 refrigerated/heating circulators for tempering double-walled reaction vessels, reaction systems, and autoclaves. With a wide temperature range of –25 to 200°C circulators can handle many applications in semi-conductor, packaging, and plastics industries. A built-in variable-speed pressure/suction pump offers powerful circulation yet operates at a quiet 55 dB(A) noise level.
PID temperature control provides excellent accuracy and stability to handle most demanding applications. Connect an external 100Ω Pt RTD temperature probe (order separately) to measure and control temperature directly in the working medium. High-quality insulation allows for fast heat-up times while reducing heat input at low temperatures. USB and RS-232 interfaces allow connection to a PC; control and monitor with the labworldsoft® laboratory software. Fluid level indicator turns LED segment red if the fluid level is too high or low.
$13,245.78 - $14,476.06CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Temperature Stability (° C) Description
Availability Pricing
IKA CBC 5 basic Refrigerated/Heating Circulator, –25 to 200°C; 115V RK-28711-79 Mfr # 4165001
COMPARE
200 ±0.02 CBC 5 basic Refrigerated/Heating Circulator, –25 to 200°C; 115V
$13,245.78CAD / Each
IKA CBC 5 control Refrigerated/Heating Circulator with Wireless Controller, –25 to 200°C; 115V RK-28711-80 Mfr # 4167001
COMPARE
200 ±0.01 CBC 5 control Refrigerated/Heating Circulator with Wireless Controller, –25 to 200°C; 115V
$14,476.06CAD / Each