Idex® Stainless Steel Sample Loops for Injection Valve Models 3725-038 and 3725i-038
Square-cut, burr-free ends provide a firm, flush connection to valve ports
  • 316 stainless steel construction
  • High pressure resistance, low permeability
A sample loop is a defined length of tubing and fittings configured to match the angle of the injection valve ports. A sample is loaded and held in the loop until injection is triggered, either automatically or manually. Stainless steel loops are supplied with unswaged fittings, so the tube can be completely bottomed in the injection port before the ferrule is swaged. The loop volumes are nominal.
$422.86 - $861.93CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Tubing Inside Diameter (ID) (mm) Tubing Outside Diameter (OD) (in)
Availability Pricing
Idex Sample Loop for 3725-038/3725i-038 Valves, SS, 5 mL, 1/8" OD x 0.080" ID; 1/EA 0201648 Mfr # 3065-019
COMPARE
5 2 0.125
$422.86CAD / Each
Idex Sample Loop for 3725-038/3725i-038 Valves, SS, 10 mL, 1/8" OD x 0.080" ID; 1/EA 0201649 Mfr # 3065-023
COMPARE
10 2 0.125
$585.67CAD / Each
Idex Sample Loop for 3725-038/3725i-038 Valves, SS, 20 mL, 1/8" OD x 0.080" ID; 1/EA 0201650 Mfr # 3065-025
COMPARE
20 2 0.125
$861.93CAD / Each